Skip to Content
Merck
All Photos(1)

Key Documents

21932

Sigma-Aldrich

5(6)-Carboxynaphthofluorescein

BioReagent, suitable for fluorescence, ≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H16O7
CAS Number:
Molecular Weight:
476.43
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Quality Level

Assay

≥90% (HPLC)

form

solid

pKa 

7.6

fluorescence

λex 512 nm; λem 567 nm (acid/neutral)
λex 598 nm; λem 668 nm (basic)

suitability

suitable for fluorescence

SMILES string

OC(=O)c1ccc2c(c1)C(=O)OC23c4ccc5cc(O)ccc5c4Oc6c3ccc7cc(O)ccc67.OC(=O)c8ccc9C(=O)OC%10(c%11ccc%12cc(O)ccc%12c%11Oc%13c%10ccc%14cc(O)ccc%13%14)c9c8

InChI

1S/2C29H16O7/c30-17-4-6-19-14(11-17)1-9-23-25(19)35-26-20-7-5-18(31)12-15(20)2-10-24(26)29(23)22-8-3-16(27(32)33)13-21(22)28(34)36-29;30-17-4-7-19-14(11-17)2-9-22-25(19)35-26-20-8-5-18(31)12-15(20)3-10-23(26)29(22)24-13-16(27(32)33)1-6-21(24)28(34)36-29/h2*1-13,30-31H,(H,32,33)

InChI key

ANUHNEJUELPITC-UHFFFAOYSA-N

Application

5(6)-Carboxynaphthofluorescein (CNF) may be used as an intravesicular pH-sensitive probe for the detection of activity, selectivity and cooperativity of ion channels such as gramicidin A.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shannon G Klein et al.
Global change biology, 23(9), 3690-3703 (2017-04-09)
Anthropogenic nutrient inputs enhance microbial respiration within many coastal ecosystems, driving concurrent hypoxia and acidification. During photosynthesis, Symbiodinium spp., the microalgal endosymbionts of cnidarians and other marine phyla, produce O
M Ramanathan et al.
Biosensors & bioelectronics, 22(12), 3001-3007 (2007-02-03)
The aim of the present work is to develop an evanescence wave array biosensor exploiting the "kinetic" approach of enzymatic reaction and further detection of the reaction products via pH sensitive fluorophore reporter. To demonstrate the feasibility of this approach
A Song et al.
Analytical chemistry, 69(5), 863-867 (1997-03-01)
A fast and durable ratiometric pH microoptode that is highly accurate, precise, sensitive, reversible, and reproducible over the physiological ranges of pH, ionic strength, and temperature has been developed. The sensing site consists of 5 (and 6)-carboxynaphthofluorescein (CNF) entrapped in
D A Jankowska et al.
Biosensors & bioelectronics, 87, 312-319 (2016-08-31)
Aging population and longer life expectancy are the main reasons for an increasing number of patients with wound problems. Although the interest in wound care increases continuously, wound management still remains a challenge mainly due to the higher occurrence of
Giorgia Giovannini et al.
Nanomaterials (Basel, Switzerland), 11(1) (2021-01-21)
The development of hybrid materials with unique optical properties has been a challenge for the creation of high-performance composites. The improved photophysical and photochemical properties observed when fluorophores interact with clay minerals, as well as the accessibility and easy handling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service