14419C
EX-CELL® 420 Serum-Free Medium for Insect Cells
Protein-free, without L-glutamine, liquid, sterile-filtered, suitable for insect cell culture
Pharma Manufacturing
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
description
for research or for further manufacturing use
sterility
sterile-filtered
form
liquid
technique(s)
cell culture | insect: suitable
components
NaHCO3: 0.35 g/L
L-glutamine: no
glucose: 6 g/L
HEPES: no
shipped in
ambient
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
EX-CELL 420 is a complete medium developed and optimized for the serum-free growth of Sf9 and Sf21 insect cell lines. Cells can be subcultured directly into EX-CELL 420 from serum-free or seum-supplemented media without adaptation. Cultures in EX-CELL 420 routinely reach cell densities greater than 1 × 107 cells/mL with greater than 95% viability. Suspension cultures can be maintained, without refeeding, for more than 10 days. Sf9 and Sf21 cells have been carried for more than 20 passages in EX-CELL 420 with no loss of viability. Protein expression and virus production are improved over serum-containing media.
Legal Information
Available in USA, Canada, and Europe. Please check with your local supplier regarding availability and restrictions in other countries.
EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany
comparable product
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cytotechnology, 43(1-3), 3-10 (2008-11-13)
The production of beta-galactosidase by Sf9 cells infected with recombinant Autographa californica nucleopolyhedrovirus (AcNPV) was investigated in shake-flask culture using two serum-free basal media: Grace's medium and TNM-FH (Grace's medium supplemented with lactalbumin hydrolysate and yeast extract). At the time
Journal of bioscience and bioengineering, 95(2), 185-187 (2005-10-20)
When rat protein kinase C-delta (PKC-delta) was produced by Sf9 cells infected with a recombinant baculovirus in shake-flask culture using a serum-containing medium, the intracellular PKC-delta content decreased in the late period while the extracellular PKC-6 markedly increased. During the
Cytotechnology, 49(1), 1-9 (2008-11-13)
Animal cells can be cultured both in basal media supplemented with fetal bovine serum (FBS) and in serum-free media. In this work, the supplementation of Grace's medium with a set of nutrients to reduce FBS requirements in Spodoptera frugiperda (Sf9)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service