137252
BIS-TRIS hydrochloride
EMPROVE® EVOLVE
Pharma Manufacturing
Synonym(s):
BIS-TRIS hydrochloride, 2,2-Bis(hydroxymethyl)-2,2′,2″-nitrilotriethanol, 2-Bis(2-hydroxyethyl)amino-2-(hydroxymethyl)-1,3-propanediol, Bis(2-hydroxyethyl)aminotris(hydroxymethyl)methane
About This Item
Recommended Products
biological source
synthetic
Quality Level
product line
EMPROVE® EVOLVE
Assay
≥99%
form
powder
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, heavy metals, trace metals, residual solvents, tested
pH
5.8-7.2
useful pH range
5.8-7.2
6.5
pKa
6.5
suitability
suitable for manufacturing use
application(s)
pharmaceutical (small molecule)
foreign activity
Cytotoxicity, DNase, NICKase, RNase, and Protease; tested
storage temp.
2-30°C
SMILES string
Cl.OCCN(CCO)C(CO)(CO)CO
InChI
1S/C8H19NO5.ClH/c10-3-1-9(2-4-11)8(5-12,6-13)7-14;/h10-14H,1-7H2;1H
InChI key
VEYRVLHAMHQVTC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service