Skip to Content
Merck
All Photos(2)

Documents

B32807

Sigma-Aldrich

Benzyltriphenylphosphonium chloride

99%

Synonym(s):

BTPPC, NSC 116712, Triphenyl(phenylmethyl)phosphonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2P(Cl)(C6H5)3
CAS Number:
Molecular Weight:
388.87
Beilstein:
3599868
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

reaction suitability

reaction type: C-C Bond Formation

mp

≥300 °C (lit.)

SMILES string

[Cl-].C(c1ccccc1)[P+](c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C25H22P.ClH/c1-5-13-22(14-6-1)21-26(23-15-7-2-8-16-23,24-17-9-3-10-18-24)25-19-11-4-12-20-25;/h1-20H,21H2;1H/q+1;/p-1

InChI key

USFRYJRPHFMVBZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Benzyltriphenylphosphonium chloride can be used as a reactant for the synthesis of:
  • Platinum chloro-tetrazole complexes via azidation.
  • Trans-stilbenes and cinnamates via Wittig olefination.
  • Achiral N-hydroxyformamide inhibitors of ADAM-TS4 and ADAM-TS5 for osteoarthritis treatment.
  • Pentiptycenes for use as light-driven molecular brakes.
  • Archipelago structures for formation of petroleum asphaltenes.


It is also used as a crosslinking agent for tube-like natural halloysite / fluorelastomer nanocomposites.
Crosslinking agent for tube-like natural halloysite / fluorelastomer nanocomposites

Reactant for synthesis of:
  • Platinum chloro tetrazole complexes via azidation
  • Trans-stilbenes and cinnamates via Wittig olefination
  • Achiral N-hydroxyformamide inhibitors of ADAM-TS4 and ADAM-TS5 for osteoarthritis treatment
  • Pentiptycenes for use as light-driven molecular brakes

Reactant for formation of archipelago structures for formation of petroleum asphaltenes

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - STOT RE 1 - STOT SE 3

Target Organs

Lungs,nasal cavity, Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

572.0 °F - closed cup

Flash Point(C)

300 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pentiptycene?Derived Light?Driven Molecular Brakes: Substituent Effects of the Brake Component.
Sun W T, et al.
Chemistry?A European Journal , 16(38), 11594-11604 (2010)
Orally active achiral N-hydroxyformamide inhibitors of ADAM-TS4 (aggrecanase-1) and ADAM-TS5 (aggrecanase-2) for the treatment of osteoarthritis.
De Savi , et al.
Bioorganic & Medicinal Chemistry Letters, 21(11), 3301-3306 (2011)
Synthesis and tyrosinase inhibition activity of trans-stilbene derivatives.
Ismail T, et al.
Bioorganic Chemistry, 64, 97-102 (2016)
Formation of archipelago structures during thermal cracking implicates a chemical mechanism for the formation of petroleum asphaltenes.
Alshareef A H, et al.
Energy and Fuels, 25(5), 2130-2136 (2011)
A new route to N (1)-5R-tetrazole complexes via azidation to nitriles coordinated to Pt (II) and Pt (IV).
Popova E A, et al.
Inorgorganica Chimica Acta, 375(1), 242-247 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service