Skip to Content
Merck
All Photos(1)

Key Documents

12010

Sigma-Aldrich

Benzaldehyde

puriss. p.a., ≥99.0% (GC)

Synonym(s):

Bitter almond

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CHO
CAS Number:
Molecular Weight:
106.12
Beilstein:
471223
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39023701
PubChem Substance ID:
NACRES:
NA.21
technique(s):
GC/GC: suitable
bp:
178-179 °C (lit.)
vapor pressure:
4 mmHg ( 45 °C)
Pricing and availability is not currently available.

vapor density

3.7 (vs air)

Quality Level

vapor pressure

4 mmHg ( 45 °C)

grade

puriss. p.a.

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

374 °F

expl. lim.

1.4 %, 20 °F

technique(s)

GC/GC: suitable

impurities

chlorine, none detected
≤0.001% nitrobenzene (C6H5NO2)
≤1% acid (as benzoic acid)

refractive index

n20/D 1.545 (lit.)
n20/D 1.545

pH

5.9 (20 °C)

bp

178-179 °C (lit.)

mp

−26 °C (lit.)

density

1.044 g/cm3 at 20 °C (lit.)

cation traces

Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Zn: ≤1 mg/kg

functional group

aldehyde
phenyl

SMILES string

O=Cc1ccccc1

InChI

1S/C7H6O/c8-6-7-4-2-1-3-5-7/h1-6H

InChI key

HUMNYLRZRPPJDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzaldehyde is the simplest aromatic aldehyde building block.

Application

Benzaldehyde has been used in:

  • Preparation of optically active 1-phenylpropan-1-ol.[1]
  • Synthesis of meso-tetraphenylporphins and chlorins.[2]

Other Notes

Sales restrictions may apply

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective addition of diethylzinc to benzaldehyde catalyzed by a small amount of chiral 2-amino-1-alcohols.
Oguni N and Omi T.
Tetrahedron Letters, 25(26), 2823-2824 (1984)
Porphyrinogens and porphodimethenes, intermediates in the synthesis of meso-tetraphenylporphins from pyrroles and benzaldehyde.
Dolphin D.
Journal of Heterocyclic Chemistry, 7, 275-283 (1970)
Yves F Widmer et al.
eLife, 7 (2018-10-23)
Lasting changes in gene expression are critical for the formation of long-term memories (LTMs), depending on the conserved CrebB transcriptional activator. While requirement of distinct neurons in defined circuits for different learning and memory phases have been studied in detail
Structure and electronic nature of the benzaldehyde/boron trifluoride adduct.
M T Reetz et al.
Journal of the American Chemical Society, 108(9), 2405-2408 (1986-04-01)
Determination of base properties of hydrotalcites: condensation of benzaldehyde with ethyl acetoacetate.
Corma A, et al.
J. Catal., 134(1), 58-65 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service