Skip to Content
Merck
All Photos(1)

Key Documents

156R-9

Sigma-Aldrich

CD56 (MRQ-42) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MRQ-42, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (156R-94)
vial of 0.5 mL concentrate (156R-95)
bottle of 1.0 mL predilute (156R-97)
vial of 1.0 mL concentrate (156R-96)
bottle of 7.0 mL predilute (156R-98)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1

control

neuroblastoma

shipped in

wet ice

storage temp.

2-8°C

visualization

membranous

Gene Information

human ... NCAM1(4684)

General description

CD56, also known as a neural cell adhesion molecule (NCAM), is a calcium-independenthomophilic binding protein that belongs to a group of cell adhesion molecules includingcadherins, selectins, and integrins. CD56 is involved in cell-cell adhesion of neural cellsduring embryogenesis and is expressed in most neuroectodermally derived tissues. Innormal tissue, anti-CD56 labels neurons, glia, Schwann cells, NK (natural killer) cells, and asubset of T-cells. CD56 expression can be seen in most NK cell neoplasms,4 and is frequentlyused to identify neoplasms with neuroendocrine differentiation.

Quality


IVD

IVD

IVD

RUO

Linkage

CD56 Positive Control Slides, Product No. 156S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Michalides et al.
International journal of cancer. Supplement = Journal international du cancer. Supplement, 8, 34-37 (1994-01-01)
The presence of the neural cell adhesion molecule, NCAM, is indicative for a poor prognosis in lung-cancer patients. Using MAb 735, we have investigated the expression of polysialic acid, PSA, on NCAM in a spectrum of neuro-endocrine lung tumors, ranging
Jianguo Tao et al.
The American journal of surgical pathology, 26(1), 111-118 (2002-01-05)
We report an unusual case of aggressive natural killer (NK)-like cytotoxic T-cell lymphoma in a previously healthy immunocompetent West African male. He presented with a fever of unknown origin, subsequently developed erythematous skin nodules, generalized lymphadenopathy, and hepatosplenomegaly, and then
Scott A Ely et al.
The American journal of pathology, 160(4), 1293-1299 (2002-04-12)
Unlike monoclonal gammopathy of undetermined significance (MGUS) or non-Hodgkin's lymphomas (NHLs) with plasmacytoid differentiation, multiple myeloma (MM) is commonly associated with lytic bone lesions. Although the mechanisms of increased osteoclast activity are partially understood, comparatively little is known about the
O Kaufmann et al.
Human pathology, 28(12), 1373-1378 (1998-01-07)
CD56 is immunohistochemically detectable in virtually all small cell carcinomas on frozen sections. The authors retrospectively tested the usefulness of the monoclonal antibody 123C3 against CD56 to differentiate pulmonary and extrapulmonary small cell carcinomas from nonneuroendocrine non-small cell carcinomas by
R E Kibbelaar et al.
European journal of cancer (Oxford, England : 1990), 27(4), 431-435 (1991-01-01)
We investigated the expression of the neural cell adhesion molecule (NCAM) in a series of surgically resected lung carcinomas of various histological subtypes by means of a panel of monoclonal antibodies recognising different N-CAM epitopes. In a subgroup of 56

Questions

1–3 of 3 Questions  
  1. Which isoforms of the CD56 antigen does the CD56 MRQ-42 clone recognize? I am curious about the 120kD, 140kD, and 180 kD isoforms, in particular.

    1 answer
    1. Unfortunately, the specific isoforms that this antibody reacts to have not been investigated. The immunogen is from the C-terminus. Please see the links below to review additional information that may be helpful:
      NP_851996
      https://www.ncbi.nlm.nih.gov/protein/NP_851996
      Entrez Gene 4684 Human
      https://www.ncbi.nlm.nih.gov/gene?cmd=Retrieve&dopt=Graphics&list_uids=4684

      Helpful?

  2. What is the stability of cut slides for staining with the CD56 MRQ-42 clone, particularly for slides that have been sectioned 6 months to 1 year ago and stored at -20°C with desiccant?

    1 answer
    1. It is generally recommended to begin with fresh cut tissue for use in IHC. When storing pre-cut tissue slides, it is typically done under refrigerated conditions (2 to 8°C), which can last for 2 to 3 years. However, the potential effects of storing at temperatures of -20°C with desiccant are not well-known in terms of tissue preservation. The best practice is to always use fresh cut tissue sections when possible.

      Helpful?

  3. Is there information on what terminus was for CD56 MRQ-42?

    1 answer
    1. Based on the information available, it seems that it is C-terminus.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service