Skip to Content
Merck
All Photos(1)

Key Documents

04259

Sigma-Aldrich

N-Ethylmaleimide

BioUltra, ≥99.0% (HPLC)

Synonym(s):

NEM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO2
CAS Number:
Molecular Weight:
125.13
Beilstein:
112448
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.26

product line

BioUltra

Quality Level

Assay

≥99.0% (HPLC)

impurities

insoluble matter, passes filter test

ign. residue

≤0.05%

bp

210 °C (lit.)

mp

43-46 °C (lit.)
44-47 °C

solubility

methanol: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

cut-off at 382 nm in methanol at 1 M

storage temp.

2-8°C

SMILES string

CCN1C(=O)C=CC1=O
CCN1C(=O)C=CC1=O

InChI

1S/C6H7NO2/c1-2-7-5(8)3-4-6(7)9/h3-4H,2H2,1H3

InChI key

HDFGOPSGAURCEO-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

N-Ethylmaleimide (NEM) has been used:
  • to prepare NEM-modified myosin beads
  • to block unmodified free thiols in samples
  • as a supplement in IP lysis/wash buffer for in vitro SUMOylation assay

Reagent for the covalent modification of cysteine residues in proteins.

Biochem/physiol Actions

Augments currents from native M-channels in sympathetic neurons and acts as an opener for KCNQ2, KCNQ4 and KCNQ5 channels.
Sulfhydryl alkylating agent that inactivates NADP-dependent isocitrate dehydrogenase and many endonucleases.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

163.2 °F - closed cup

Flash Point(C)

72.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measuring the kinetic and mechanical properties of non-processive myosins using optical tweezers
Greenberg MJ, et al.
Methods in Molecular Biology, 483-509 (2017)
SUMOylation of periplakin is critical for efficient reorganization of keratin filament network
Gujrati M, et al.
Molecular Biology of the Cell, 30(3), 357-369 (2019)
Resin-assisted enrichment of thiols as a general strategy for proteomic profiling of cysteine-based reversible modifications
Jia Guo
Nature Protocols, 9(1), 64-75 (2014)
Dar'ya S Redka et al.
Biochemistry, 52(42), 7405-7427 (2013-09-21)
Muscarinic and other G protein-coupled receptors exhibit an agonist-specific heterogeneity that tracks efficacy and commonly is attributed to an effect of the G protein on an otherwise homogeneous population of sites. To examine this notion, M2 muscarinic receptors were purified
Tatsuki Kurokawa et al.
Biochimica et biophysica acta, 1838(1 Pt B), 382-387 (2013-10-22)
Hv1 (also named, voltage-sensor only protein, VSOP) lacks an authentic pore domain, and its voltage sensor domain plays both roles in voltage sensing and proton permeation. The activities of a proton channel are intrinsic to protomers of Hv1, while Hv1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service