Skip to Content
Merck
All Photos(3)

Key Documents

M51008

Sigma-Aldrich

Methyliminodiacetic acid

99%

Synonym(s):

MIDA, MIDA ligand

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3N(CH2CO2H)2
CAS Number:
Molecular Weight:
147.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

220 °C (dec.) (lit.)

SMILES string

CN(CC(O)=O)CC(O)=O

InChI

1S/C5H9NO4/c1-6(2-4(7)8)3-5(9)10/h2-3H2,1H3,(H,7,8)(H,9,10)

InChI key

XWSGEVNYFYKXCP-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mengping Guo et al.
Organic & biomolecular chemistry, 12(36), 7136-7139 (2014-08-12)
N-Methyliminodiacetic acid (MIDA) as a simple, air stable and water-soluble ligand has been used in the palladium-catalyzed Hiyama cross-coupling reaction of trimethoxyphenylsilane with aryl halides. The yield of the corresponding Hiyama coupling products is high up to around 90% in
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 25(10), 1144-1149 (1984-10-01)
A new approach to radiopharmaceutical design is demonstrated, in which small chelating groups capable of binding gamma-emitting radiometals are attached to biologically active molecules, thus producing radiopharmaceuticals based on bifunctional drug and biochemical analogs. The chelating group iminodiacetic acid has
Juan Lorenzo Hagad et al.
Sensors (Basel, Switzerland), 21(5) (2021-04-04)
Two of the biggest challenges in building models for detecting emotions from electroencephalography (EEG) devices are the relatively small amount of labeled samples and the strong variability of signal feature distributions between different subjects. In this study, we propose a
Gas-liquid chromatography and enzymatic determination of alanopine and strombine in tissues of marine invertebrates.
K B Storey et al.
Analytical biochemistry, 125(1), 50-58 (1982-09-01)
G B Fiore et al.
Analytical biochemistry, 139(2), 413-417 (1984-06-01)
A method for the separation and quantification of the levels of alanopine and strombine in neutralized, perchloric acid extracts of tissues of marine invertebrates is presented. The method is based on high-performance liquid chromatographic (HPLC) separation, postcolumn derivatization using o-phthaldialdehyde

Articles

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service