Skip to Content
Merck
All Photos(1)

Key Documents

T59803

Sigma-Aldrich

Triethylene glycol dimethyl ether

ReagentPlus®, 99%

Synonym(s):

1,2-Bis(2-methoxyethoxy)ethane, Dimethyltriglycol, NSC 66400, Triglyme

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2CH2O)3CH3
CAS Number:
Molecular Weight:
178.23
Beilstein:
1700630
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21
Assay:
99%
bp:
216 °C (lit.)
Pricing and availability is not currently available.

vapor density

>4.7 (vs air)

Quality Level

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

375 °F

expl. lim.

8 %

refractive index

n20/D 1.423 (lit.)

bp

216 °C (lit.)

mp

−45 °C (lit.)

density

0.986 g/mL at 25 °C (lit.)

SMILES string

COCCOCCOCCOC

InChI

1S/C8H18O4/c1-9-3-5-11-7-8-12-6-4-10-2/h3-8H2,1-2H3

InChI key

YFNKIDBQEZZDLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethyleneglycol dimethyl ether, also known as triglyme, is a member of the glycol ether family and is commonly used as a solvent in industries like electronics, pharmaceuticals, coatings, and more. Triglymes are chemically inert because they lack reactive functional groups, making them well-suited for chemical synthesis. They have strong solvating abilities and demonstrate excellent thermal and chemical stability, which makes them highly valuable as solvents and processing aids in the manufacturing and formulation of industrial chemicals.

Application

Triethylene glycol dimethyl ether can be used as a:
  • Extracting solvent in a modified SA-HPSW (Supercritical Anti-Solvent with High-Pressure Solvent Wash) system.
  • Solvent in the synthesis of passivated iron nanoparticles.[1]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of tetraethylene glycol passivated iron nanoparticles
da Silva Nunes E, et al.
Applied Surface Science, 315, 337-345 (2014)
H Liu et al.
Dalton transactions (Cambridge, England : 2003), 46(33), 10986-10995 (2017-08-05)
This work reports two new silver metalorganic precursors for the chemical vapor deposition of Ag metallic coatings. Both precursors are based on β-diketonate adducts, namely, Ag(hfac)(L) (H-hfac = 1,1,1,5,5,5-hexafluoro-2,4-pentanedione), where L is 1,10-phenanthroline (phen) or 2,5,8,11-tetraoxadodecane (triglyme). Using these ligands
N L Bossert et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 18(4), 602-608 (1992-05-11)
Triethylene glycol and two of its derivatives were evaluated for reproductive toxicity in a continuous breeding protocol with Swiss CD-1 mice. Triethylene glycol (TEG: 0, 0.3, 1.5, and 3%), triethylene glycol diacetate (TGD: 0, 0.75, 1.5, and 3%), and triethylene
J D George et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 9(1), 173-181 (1987-07-01)
Triethylene glycol dimethyl ether (triEGdiME) is structurally related to several compounds which produce reproductive and developmental toxicity, including teratogenicity in laboratory animals. In the present study, triEGdiME (0, 250, 500, or 1000 mg/kg/day) was administered by gavage to timed-pregnant CD-1
Dilek Cökeliler et al.
Dental materials : official publication of the Academy of Dental Materials, 23(3), 335-342 (2006-05-17)
This study evaluates the effect of plasma treated E-glass fiber to improve the mechanical properties of acrylic resin denture base material, polymethylmethacrlyate (PMMA). Plasma surface treatment of fibers is used as reinforcement in composite materials to modify the chemical and

Questions

  1. こちらの商品(T59803 トリエチレングリコールジメチルエーテル)には酸化防止剤(BHT等)は入っていますか?入っている場合濃度はどのくらいですか?

    1 answer
    1. Product No. T59803 (Triethylene glycol dimethyl ether, ReagentPlus®, 99%) does not contain any antioxidants or inhibitors. It consists of neat triethylene glycol dimethyl ether with a specified GC purity ≥ 98.5 %. The actual lot-specific purity often is higher than this value, but the residual impurities in this product are not identified.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service