Skip to Content
Merck
All Photos(4)

Documents

900502

Sigma-Aldrich

Potassium carbonate

anhydrous, free-flowing, −325 mesh, Redi-Dri, reagent grade, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2CO3
CAS Number:
Molecular Weight:
138.21
Beilstein:
4267587
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous
reagent grade

Quality Level

product line

Redi-Dri

Assay

≥98%

form

powder

quality

free-flowing

particle size

−325 mesh

pH

11-13 (25 °C, 138 g/L)

mp

891 °C (lit.)

SMILES string

[K+].[K+].[O-]C([O-])=O

InChI

1S/CH2O3.2K/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

BWHMMNNQKKPAPP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Potassium carbonate is an inorganic base. It participates as base in the palladacycle catalyzed Heck reaction of chlorobenzene with styrene.

Application

Potassium carbonate was used in the following studies:
  • Synthesis of polysubstituted iodobenzene derivatives.
  • As base for the synthesis of high molecular weight homopolymers and copolymers derived from various bisphenols.
  • As base for the Suzuki coupling of aryl halides with aryl boronic acids.
  • As base for the Heck reaction of styrene and bromobenzene.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, kinetic observations and characteristics of polyarylene ether sulphones prepared via a potassium carbonate DMAC process.
Viswanathan R, et al.
Polymer, 25(12), 1827-1836 (1984)
Robert Möckel et al.
Organic letters, 17(7), 1644-1647 (2015-03-21)
The cobalt-catalyzed Diels-Alder reaction of trimethylsilyl-substituted alkynes with 1,3-dienes led to dihydroaromatic intermediates which were transformed into iodobenzene derivatives. For this transformation, the dihydroaromatic intermediates had to be oxidized and the trimethylsilyl-substituted arene had to undergo a silicon-iodine exchange reaction.
Chelating N-heterocyclic carbene ligands in palladium-catalyzed Heck-type reactions.
Herrmann WA, et al.
Journal of Organometallic Chemistry, 557(1), 93-96 (1998)
Heck reaction using palladium complexed to dendrimers on silica.
Alper H, et al.
Canadian Journal of Chemistry, 78(6), 920-924 (2000)
Carlos Arroniz et al.
Organic letters, 15(4), 910-913 (2013-02-05)
ortho-Arylation of ortho-substituted benzoic acids is a challenging process due to the tendency of the reaction products toward Pd-catalyzed protodecarboxylation. A simple method for preventing decarboxylation in sterically hindered benzoic acids is reported. The method described represents a reliable and

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service