Skip to Content
Merck
All Photos(3)

Key Documents

900501

Sigma-Aldrich

Potassium carbonate

anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2CO3
CAS Number:
Molecular Weight:
138.21
Beilstein:
4267587
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
99%
grade:
anhydrous
form:
powder
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

product line

ReagentPlus®
Redi-Dri

Assay

99%

form

powder

quality

free-flowing

pH

11-13 (25 °C, 138 g/L)

mp

891 °C (lit.)

SMILES string

[K+].[K+].[O-]C([O-])=O

InChI

1S/CH2O3.2K/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

BWHMMNNQKKPAPP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Improved 2-pyridyl reductive homocoupling reaction using biorenewable solvent: A detailed examination of using potassium carbonate in an enhanced reductive homocoupling of 2-pyridyl compounds in a biorenewable solvent, showcasing a sustainable approach to chemical synthesis (Webb et al., 2023).

  • Fe(3)O(4)@void@C-Schiff-base/Pd yolk-shell nanostructures as a nanocatalyst for Suzuki coupling reaction: Discusses the development of a palladium-based nanocatalyst supported by potassium carbonate for improved Suzuki coupling reactions, an important reaction in organic chemistry (Barzkar et al., 2023).

  • Enhanced LC-ESI-MS/MS Sensitivity by Cationic Derivatization of Organophosphorus Acids: Study on enhancing liquid chromatography-electrospray ionization tandem mass spectrometry sensitivity using potassium carbonate, improving the detection of organophosphorus acids (Shamai Yamin et al., 2023).


Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chelating N-heterocyclic carbene ligands in palladium-catalyzed Heck-type reactions.
Herrmann WA, et al.
Journal of Organometallic Chemistry, 557(1), 93-96 (1998)
Robert Möckel et al.
Organic letters, 17(7), 1644-1647 (2015-03-21)
The cobalt-catalyzed Diels-Alder reaction of trimethylsilyl-substituted alkynes with 1,3-dienes led to dihydroaromatic intermediates which were transformed into iodobenzene derivatives. For this transformation, the dihydroaromatic intermediates had to be oxidized and the trimethylsilyl-substituted arene had to undergo a silicon-iodine exchange reaction.
Heck reaction using palladium complexed to dendrimers on silica.
Alper H, et al.
Canadian Journal of Chemistry, 78(6), 920-924 (2000)
Polyamide interfacial composite membranes prepared from m-phenylene diamine, trimesoyl chloride and a new disulfonated diamine.
Xie W, et al.
Journal of Membrane Science, 403, 152-161 (2012)
Carlos Arroniz et al.
Organic letters, 15(4), 910-913 (2013-02-05)
ortho-Arylation of ortho-substituted benzoic acids is a challenging process due to the tendency of the reaction products toward Pd-catalyzed protodecarboxylation. A simple method for preventing decarboxylation in sterically hindered benzoic acids is reported. The method described represents a reliable and

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service