Skip to Content
Merck
All Photos(1)

Key Documents

71702

Sigma-Aldrich

Sodium iodate

puriss. p.a., ≥99.5% (RT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaIO3
CAS Number:
Molecular Weight:
197.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

puriss. p.a.

Assay

≥99.5% (RT)

form

powder or crystals

impurities

≤0.005% total nitrogen (N)

pH

5.0-8.0 (25 °C, 50 mg/mL in H2O)

solubility

water: soluble(lit.)

density

4.28 g/mL at 25 °C (lit.)

anion traces

bromide, bromate, chloride, chlorate (as Cl-): ≤200 mg/kg
iodide (I-): ≤20 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤100 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤10 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[Na+].[O-]I(=O)=O

InChI

1S/HIO3.Na/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

WTCBONOLBHEDIL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium iodate (NaIO3) is colorless sodium salt having rhombic crystals. It exhibits oxidizing properties. In nature, it is found in Chile saltpeter.

Application

Sodium iodate may be employed as a starting reagent in the preparation of iodine and its derivatives. It may be employed as a catalyst in the synthesis of bis-indol.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Ox. Sol. 2 - Skin Sens. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bis (indolyl) methanes Synthesis Through Sodium Iodate and Sodium Hydrogen Sulfite in Water.
Mohammed Khan K, et al.
J. Chem. Soc. Pak., 36(6) (2014)
Alyson Wolk et al.
Redox biology, 37, 101681-101681 (2020-08-24)
Sorsby Fundus Dystrophy (SFD) is a rare inherited autosomal dominant macular degeneration caused by specific mutations in TIMP3. Patients with SFD present with pathophysiology similar to the more common Age-related Macular Degeneration (AMD) and loss of vision due to both
Luisa M Franco et al.
Investigative ophthalmology & visual science, 50(8), 4004-4010 (2009-04-03)
To correlate damage to the retinal pigment epithelium (RPE) with decreased visual function after the systemic administration of sodium iodate (NaIO(3)). Damage was produced in mice by injection of 15, 25, or 35 mg/kg NaIO(3). Visual function was assessed with
William S Redfern et al.
Journal of pharmacological and toxicological methods, 63(1), 102-114 (2010-07-14)
We have evaluated the ability of a semi-automated, optomotor reflex method to assess drug-induced visual dysfunction, in albino and pigmented rats and mice. Male Han Wistar (HW) and Long Evans (LE) rats and mice (CD-1 and C57BL/6) were tested in
Huiming Xia et al.
Experimental eye research, 93(6), 862-872 (2011-10-13)
Regenerative medicine holds the promise of restoring cells and tissues that are destroyed in human disease, including degenerative eye disorders. However, development of this approach in the eye has been limited by a lack of animal models that show robust

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service