Skip to Content
Merck
All Photos(1)

Key Documents

676926

Sigma-Aldrich

2-Butanone

ACS reagent, ≥99.0%

Synonym(s):

Ethyl methyl ketone, MEK, Methyl ethyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5COCH3
CAS Number:
Molecular Weight:
72.11
Beilstein:
741880
EC Number:
MDL number:
UNSPSC Code:
12352115
eCl@ss:
39021202
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
bp:
80 °C (lit.)
vapor pressure:
71 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor density

2.49 (vs air)

vapor pressure

71 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

960 °F

expl. lim.

10.1 %

dilution

(for analytical testing)

impurities

≤0.0005 meq/g Titr. acid
≤0.20% water

evapn. residue

≤0.0025%

color

APHA: ≤15

refractive index

n20/D 1.379 (lit.)

bp

80 °C (lit.)

mp

−87 °C (lit.)

density

0.805 g/mL at 25 °C (lit.)

SMILES string

CC(CC)=O

InChI

1S/C4H8O/c1-3-4(2)5/h3H2,1-2H3

InChI key

ZWEHNKRNPOVVGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Butanone is a ketone. Physical properties of the solutions of 1-n-butyl-3-methylimidazolium tetrafluoroborate (ionic liquid) in various organic solvents (acetonitrile, dichloromethane, 2-butanone and N,N-dimethylformamide) has been studied. Viscosity and density of these mixtures were evaluated at 298.15K.[1]

Application

2-Butanone (Methyl ethyl ketone) may be employed as solvent for the methylation agent [11C]methyl trifluoromethanesulfonate ([11C]CH3OTf), during the completely automated radiosynthesis of 2-(4-N-[11C]methylaminophenyl)-6-hydroxybenzothiazole ([11C]PIB).[2] It may be used in the synthesis of nanocontainer composed of 6 bowl-shaped cavitands. In the nanocontainer, cavitands were joined by 12 -CH=N-CH2CH2-N=CH- linkers.[3]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

C Philippe et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 69(9), 1212-1217 (2011-05-10)
[(11)C]PIB is still the standard PET compound for Alzheimer imaging targeting beta-amyloid plaques. We aimed to establish a fully-automated procedure for the synthesis and purification of [(11)C]PIB with a high degree of reliability and improved specific activity as well as
A simple one-pot multicomponent synthesis of an octahedral nanocontainer molecule.
Liu X and Warmuth R.
Nature Protocols, 2(5), 1288-1296 (2007)
A volumetric and viscosity study for the mixtures of 1-n-butyl-3-methylimidazolium tetrafluoroborate ionic liquid with acetonitrile, dichloromethane, 2-butanone and N, N-dimethylformamide.
Wang J, et al.
Green Chemistry, 5(5), 618-622 (2003)
L B Goldsmith et al.
Contact dermatitis, 19(5), 348-350 (1988-11-01)
Irritant contact dermatitis along with an increased transepidermal water loss can result from exposing the skin to solvents. A study of the interaction of various solvents with human stratum corneum was made using thin-layer chromatography. Comparison of 10 solvents (trichloroethylene
S G Carmella et al.
Cancer research, 53(4), 721-724 (1993-02-15)
Metabolites of the tobacco-specific nitrosamine, 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone, a potent pulmonary carcinogen, have been quantified in the urine of 11 smokers. They were not detected in nonsmokers' urine. The metabolites, 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanol and its glucuronide, were detected in quantities of 0.23-1.0 and 0.57-6.5

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service