Skip to Content
Merck
All Photos(1)

Key Documents

673803

Sigma-Aldrich

Diisopropyl ether

contains either BHT or hydroquinone as stabilizer, ACS reagent, ≥99.0%

Synonym(s):

Isopropyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOCH(CH3)2
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731256
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.07
grade:
ACS reagent
Assay:
≥99.0%
bp:
68-69 °C (lit.)
vapor pressure:
120 mmHg ( 20 °C)

grade

ACS reagent

Quality Level

vapor density

3.5 (vs air)

vapor pressure

120 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

827 °F

contains

either BHT or hydroquinone as stabilizer

expl. lim.

1-21 %, 100 °F

impurities

≤0.0007 meq/g Titr. acid
≤0.05% peroxide (as C6H14O2)

evapn. residue

≤0.01%

color

APHA: ≤25

refractive index

n20/D 1.367 (lit.)

bp

68-69 °C (lit.)

mp

−85 °C (lit.)

density

0.725 g/mL at 25 °C (lit.)

SMILES string

CC(C)OC(C)C

InChI

1S/C6H14O/c1-5(2)7-6(3)4/h5-6H,1-4H3

InChI key

ZAFNJMIOTHYJRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-20.2 °F - closed cup

Flash Point(C)

-29 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B E Cham et al.
Journal of virological methods, 137(1), 160-163 (2006-07-01)
Many viruses including HIV, hepatitis C and hepatitis B, have an outer lipid envelope which maintains inserted viral peptides in the "correct" functional conformation and orientation. Disruption of the lipid envelope by most solvents destroys infectivity and often results in
R Gonzalez-Olmos et al.
Chemosphere, 71(11), 2098-2105 (2008-02-27)
Methyl tert-butyl ether (MTBE) is the most widely used oxygenate in gasoline blending and has become one of the world's most widespread groundwater and surface water pollutants. Alternative oxygenates to MTBE, namely ethyl tert-butyl ether (ETBE), tert-amyl ether (TAME) and
R van Gijn et al.
Journal of chromatography. B, Biomedical applications, 667(2), 269-276 (1995-05-19)
The staurosporine derivative, N-benzoylstaurosporine (CGP 41 251; I), is a protein kinase C inhibitor that has been selected for phase I clinical evaluation in cancer patients. We have developed a selective and sensitive assay of the drug and three potential
P Adlercreutz
Biochimica et biophysica acta, 1163(2), 144-148 (1993-05-13)
Horse liver alcohol dehydrogenase and alpha-chymotrypsin were deposited on a porous support material, Celite. After equilibration at a well-defined water activity, the catalytic activity was measured with diisopropyl ether as reaction medium. The effects of the presence of polyols and
A Hickel et al.
Biotechnology and bioengineering, 65(4), 425-436 (1999-10-03)
A novel recycle reactor has been designed to determine the interfacial activity of hydroxynitrile lyase in a diisopropyl ether (DIPE)/water two-phase system. The reactor provides a known interfacial area. Enzyme activity toward mandelonitrile cleavage is continuously measured in the reactor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service