Skip to Content
Merck
All Photos(1)

Documents

111333

Sigma-Aldrich

Dipropyl ether

≥99%

Synonym(s):

Propyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)2O
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731312
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

55 mmHg ( 20 °C)

Quality Level

Assay

≥99%

refractive index

n20/D 1.380 (lit.)

bp

88-90 °C (lit.)

mp

−123 °C (lit.)

solubility

H2O: slightly soluble
alcohols: soluble
diethyl ether: soluble

density

0.736 g/mL at 25 °C (lit.)

SMILES string

CCCOCCC

InChI

1S/C6H14O/c1-3-5-7-6-4-2/h3-6H2,1-2H3

InChI key

POLCUAVZOMRGSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dipropyl ether was used to study the low proton-transporting ability of a water molecule in the excited-state proton transfer (ESPT) of a 7-azaindole (7AI) moiety.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoko Nunome et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 36(2), 269-276 (2019-10-16)
This study describes a direct analysis of oxygenated organic compounds, such as ketones, esters and ethers, rapidly and easily using a soft plasma ionization (SPI) source combined with a Q-mass spectrometer. A related molecular ion, [2M+H]+, in which a sample
G D Nielsen et al.
Acta pharmacologica et toxicologica, 56(2), 158-164 (1985-02-01)
The sensory irritating response of propyl ether, measured as a decrease in respiratory rate in mice, faded within one minute. The threshold was 620 p.p.m., calculated from the concentration-response curve, which had a low slope. The sensory irritating level depressing
Luisa M Figueiredo et al.
Nucleic acids research, 33(3), 1111-1122 (2005-02-22)
Telomerase replicates chromosome ends, a function necessary for maintaining genome integrity. We have identified the gene that encodes the catalytic reverse transcriptase (RT) component of this enzyme in the malaria parasite Plasmodium falciparum (PfTERT) as well as the orthologous genes
M Srividhya et al.
International journal of pharmaceutics, 326(1-2), 119-127 (2006-08-26)
Amphiphilic comb like macromonomer containing hydrophilic poly(ethylene glycol) groups covalently linked to poly(hydromethyl siloxane) (PHMS) were prepared by hydrosilylation reaction. The epoxy reacting sites were introduced to this amphiphilic system by the reaction with allyl epoxy propyl ether (AEPE). Bovine
Lin Mei et al.
Nanoscale research letters, 4(12), 1530-1539 (2009-01-01)
Multidrug resistance (MDR) in tumor cells is a significant obstacle to the success of chemotherapy in many cancers. The purpose of this research is to test the possibility of docetaxel-loaded poly (ε-caprolactone)/Pluronic F68 (PCL/Pluronic F68) nanoparticles to overcome MDR in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service