Skip to Content
Merck
All Photos(1)

Key Documents

59300

Sigma-Aldrich

2-Propanol

≥99.8% (GC), ACS reagent

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.05
Assay:
≥99.8% (GC)
bp:
82 °C (lit.)
vapor pressure:
33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Product Name

2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)

grade

ACS reagent
puriss. p.a.

Quality Level

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

impurities

aldehyde, in accordance
substances darkened by H2SO4, in accordance
≤0.00017% free alkali (as NH3)
≤0.002% free acid (as C2H5COOH)
≤0.1% water

evapn. residue

≤0.001%

refractive index

n20/D 1.377 (lit.)
n20/D 1.378

pH

7.0 (20 °C)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

density

0.785 g/mL at 20 °C
0.785 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤1 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CC(C)O

storage temp.

room temp

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Propanol (Isopropanol) is a secondary alcohol. It has been tested as a substitute to fuel for use in various fuel cells. CuO powder dissolved in 2-propanol has been used for the laser ablation assisted synthesis of Cu colloids. Suspension of 2-propanol with Zn(NO3)2 has been employed for the fabrication of titanium dioxide-coated stainless steel (P25-TiO2/SS) photoanode coated with uniformly thick layer of P25-TiO2. This photoanode was used for the electrochemical photocatalytic (ECPC) degradation process.

Other Notes

The article number 59300-4X2.5L will be discontinued. Please order the single bottle 59300-2.5L which is physically identical with the same exact specifications.
The article number 59300-6X1L will be discontinued. Please order the single bottle 59300-1L which is physically identical with the same exact specifications.
Sales restrictions may apply

greener alternative product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Layered shaped alginate hydrogels for soft tissue engineering based on chemical control of the crosslinking rate.
Dimonie D, et al.
Digest Journal of Nanomaterials and Biostructures, 8(1) (2013)
A direct 2-propanol polymer electrolyte fuel cell.
Cao D and Bergens SH.
Journal of Power Sources, 124(1), 12-17 (2003)
Impact of three different TiO 2 morphologies on hydrogen evolution by methanol assisted water splitting: nanoparticles, nanotubes and aerogels
D?Elia D, et al.
International Journal of Hydrogen Energy, 36(22), 14360-14373 (2011)
Electrochemical photocatalytic degradation of dye solution with a TiO2-coated stainless steel electrode prepared by electrophoretic deposition.
Lin WC, et al.
Applied Catalysis. B, Environmental, 140, 32-41 (2013)
Evaluation of Ethanol, 1-Propanol, and 2-Propanol in a Direct Oxidation Polymer-Electrolyte Fuel Cell A Real-Time Mass Spectrometry Study.
Wang J, et al.
Journal of the Electrochemical Society, 142(12), Wang J-Wang J (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service