Skip to Content
Merck
All Photos(1)

Documents

439207

Sigma-Aldrich

2-Propanol

99.5%, suitable for HPLC

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.05

product name

2-Propanol, suitable for HPLC, 99.5%

vapor density

2.1 (vs air)

Quality Level

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Assay

99.5%

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

technique(s)

HPLC: suitable

impurities

≤0.05% water

evapn. residue

<0.0003%

refractive index

n20/D 1.377 (lit.)

pH

7.0 (20 °C)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

solubility

water: soluble

density

0.785 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 205 nm Amax: 1.0
λ: 220 nm Amax: 0.30
λ: 230 nm Amax: 0.15
λ: 245 nm Amax: 0.05
λ: 260 nm Amax: 0.03
λ: 300-400 nm Amax: 0.01

application(s)

food and beverages

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Propanol (i-PrOH), a renewable secondary alcohol, is water soluble, colorless, volatile liquid with a sweet odor. It is a potential substitute to petroleum-derived gasoline. Preparation of methyl isobutyl ketone (MIBK) using 2-propanol as the precursor has been reported with yield as high as 25%. Solubility of benzoic acid has been investigated in 2-propanol by gravimetric method in the temperature range, 277-346K. Its photocatalytic degradation has been examined by various methods.

Application

2-Propanol may be used in the following processes:
  • As a solvent in the synthesis of di- and trisubstituted ortho biaryls by Suzuki-Miyaura cross-coupling reaction.
  • As an alternate solvent to benzene in the analysis of total carbonyl compounds in heated and frying oils.
  • As a reaction medium in the preparation of cyclic ureas by reacting CO2 with diamines in the presence of pure cerium oxide (CeO2).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A general method for the Suzuki-Miyaura cross-coupling of sterically hindered aryl chlorides: Synthesis of di-and tri-ortho-substituted biaryls in 2-propanol at room temperature.
Navarro O, et al.
Journal of the American Chemical Society, 125(52), 16194-16195 (2003)
A modified method for the estimation of total carbonyl compounds in heated and frying oils using 2-propanol as a solvent.
Endo Y, et al.
Journal of the American Oil Chemists' Society, 78(10), 1021-1024 (2001)
Photocatalytic degradation of 2-propanol and phenol using Au loaded MnWO4 nanorod under visible light irradiation.
Chakraborty AK, et al.
Journal of Cluster Science, 23(2), 437-448 (2012)
Photocatalytic degradation of 2-propanol under irradiation of visible light by nanocrystalline titanium (IV) oxide modified with rhodium ion using adsorption method.
Kitano S, et al.
Chemistry Letters (Jpn), 39(6), 627-629 (2010)
Highly efficient synthesis of cyclic ureas from CO2 and diamines by a pure CeO2 catalyst using a 2-propanol solvent.
Tamura M, et al.
Green Chemistry, 15(6), 1567-1577 (2013)

Articles

UHPLC separation of Vitamin D Metabolites using Supel Carbon LC column ensures excellent peak shape and sensitivity.

UHPLC separation of Vitamin D Metabolites using Supel Carbon LC column ensures excellent peak shape and sensitivity.

UHPLC separation of Vitamin D Metabolites using Supel Carbon LC column ensures excellent peak shape and sensitivity.

UHPLC separation of Vitamin D Metabolites using Supel Carbon LC column ensures excellent peak shape and sensitivity.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service