Skip to Content
Merck
All Photos(4)

Key Documents

410241

Sigma-Aldrich

Sodium perchlorate

ACS reagent, ≥98.0%

Synonym(s):

Hyperchloric acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaClO4
CAS Number:
Molecular Weight:
122.44
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98.0%
grade:
ACS reagent
form:
powder or chunks
solubility:
water: soluble 2096 g/L at 20 °C
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Assay

≥98.0%

form

powder or chunks

reaction suitability

reagent type: oxidant

impurities

≤0.005% insolubles

pH

6.0-8.0 (25 °C, 5%)

mp

468 °C (lit.)

solubility

water: soluble 2096 g/L at 20 °C

anion traces

chloride (Cl-): ≤0.003%
sulfate (SO42-): ≤0.002%

cation traces

Ca: ≤0.02%
Fe: ≤5 ppm
K: ≤0.05%
heavy metals: ≤5 ppm (by ICP)

SMILES string

[Na+].[O-]Cl(=O)(=O)=O

InChI

1S/ClHO4.Na/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

BAZAXWOYCMUHIX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium perchlorate is a water soluble perchlorate salt. It is prepared by electrolyzing aqueous sodium chlorate solution. It is a strong oxidizing agent.[1] On impregnating porous silicon with sodium perchlorate it forms an explosive composite which has been investigated.[2]

Application


  • Silica-Poly(Vinyl Alcohol) Composite Aerogel: A Promising Electrolyte for Solid-State Sodium Batteries.: This research highlights the use of sodium perchlorate in developing silica-poly(vinyl alcohol) composite aerogels, serving as an innovative electrolyte for solid-state sodium batteries. This study showcases the potential for advancements in energy storage technologies, emphasizing the role of sodium perchlorate in enhancing battery performance and safety (Vareda JP et al., 2024).


  • A sodium ion conducting gel polymer electrolyte with counterbalance between 1-ethyl-3-methylimidazolium tetrafluoroborate and tetra ethylene glycol dimethyl ether for electrochemical applications.: This research demonstrates the use of sodium perchlorate in developing a gel polymer electrolyte for sodium-ion batteries, providing a balance between conductivity and stability, crucial for the next generation of energy storage systems (Patel M et al., 2024).



  • Bendable quasi-solid-state aqueous sodium-ion batteries operated at -30 °C.: Utilizing sodium perchlorate, this study develops flexible and low-temperature operable sodium-ion batteries, offering significant advancements in portable and wearable electronic devices, highlighting the adaptability and efficiency of sodium perchlorate in extreme conditions (Gui Q et al., 2024).


Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 1 - STOT RE 2

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Changyong Zhang et al.
Water research, 145, 220-230 (2018-08-25)
Ammonia nitrogen removal from wastewaters has gained much attention in recent decades as a result of the environmental problems associated with discharge of excessive amounts of this critical nutrient including eutrophication of receiving waters, generation of offensive odours as a
Qiufang Liu et al.
Theranostics, 8(3), 785-799 (2018-01-19)
Purpose: Transcatheter hepatic artery embolization therapy is a minimally invasive alternative for treating inoperable liver cancer but recurrence is frequent. Multifunctional agents, however, offer an opportunity for tumor eradication. In this study, we were aim to synthesized poly (lactic-co-glycolic acid)
Ajar Kamal et al.
The Analyst, 142(10), 1827-1834 (2017-05-06)
The present work describes the effect of the number of thymine-thymine mispairs in single strand DNA probes on Hg(ii) interactions and further to develop a highly sensitive DNA based impedimetric sensor for Hg(ii) detection. To achieve this goal, the influence
Eagleson M.
Concise Encyclopedia Chemistry, 1000-1000 (1994)
Synthesis of sulfones using sodium perchlorate as a catalyst under neutral conditions.
Bandgar BP, et al.
New. J. Chem., 26(9), 1105-1107 (2002)

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Do you have it on pharmaceutical grade, suitable for pharmacceutical formulations?

    1 answer
    1. This product is intended for research use only. Currently, there is no product option available that is suitable for pharmaceutical use.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service