If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf
1.06564
Sodium perchlorate monohydrate
for analysis EMSURE®
Synonym(s):
Sodium perchlorate monohydrate, Perchloric Acid Sodium Salt
About This Item
Recommended Products
Quality Level
product line
EMSURE®
Assay
99.0-102.0% (alkalimetric)
form
crystals
pH
4.5-7.0 (25 °C, 50 g/L in H2O)
mp
130 °C
solubility
2090 g/L
density
2.02 g/cm3 at 20 °C
bulk density
900 kg/m3
anion traces
chloride (Cl-): ≤0.002%
chloride, chlorate (as Cl-): ≤0.002%
sulfate (SO42-): ≤0.002%
cation traces
Ca: ≤0.002%
Fe: ≤0.0003%
K: ≤0.005%
N: ≤0.0005%
heavy metals (as Pb): ≤0.0005%
storage temp.
2-30°C
SMILES string
[Na+].[Cl](=O)(=O)(=O)[O-].O
InChI
1S/ClHO4.Na.H2O/c2-1(3,4)5;;/h(H,2,3,4,5);;1H2/q;+1;/p-1
InChI key
IXGNPUSUVRTQGW-UHFFFAOYSA-M
Linkage
Analysis Note
pH-value (5 %; water): 4.5 - 7.0
Chloride, Chlorate (as Cl): ≤ 0.002 %
Chloride (Cl): ≤ 0.002 %
Sulfate (SO₄): ≤ 0.002 %
Total nitrogen (N): ≤ 0.0005 %
Heavy metals (as Pb): ≤ 0.0005 %
Ca (Calcium): ≤ 0.002 %
Fe (Iron): ≤ 0.0003 %
K (Potassium): ≤ 0.005 %
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 1
Storage Class Code
5.1A - Strongly oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
Folic acid vs. folate differences and biological activities; implications in fortified foods and supplements for human health.
Folic acid vs. folate differences and biological activities; implications in fortified foods and supplements for human health.
Folic acid vs. folate differences and biological activities; implications in fortified foods and supplements for human health.
Folic acid vs. folate differences and biological activities; implications in fortified foods and supplements for human health.
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service