Skip to Content
Merck
All Photos(3)

Key Documents

34857

Sigma-Aldrich

1,4-Dioxane

suitable for HPLC, ≥99.5%

Synonym(s):

Diethylene oxide, Dioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
Beilstein:
102551
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.5%
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
100-102 °C (lit.)
vapor pressure:
27 mmHg ( 20 °C)
40 mmHg ( 25 °C)

vapor density

3 (vs air)

Quality Level

vapor pressure

27 mmHg ( 20 °C)
40 mmHg ( 25 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

356 °F

expl. lim.

22 %

technique(s)

HPLC: suitable

impurities

≤0.001% non-volatile matter
≤0.002% free acid (as CH3COOH)
≤0.01% peroxides (as H2O2)
≤0.05% water (Karl Fischer)

transmittance

220 nm, ≥20%
270 nm, ≥80%
300 nm, ≥98%

refractive index

n20/D 1.422 (lit.)

pH

6.0-8 (20 °C, 500 g/L)

bp

100-102 °C (lit.)

mp

10-12 °C (lit.)

density

1.034 g/mL at 25 °C (lit.)

UV absorption

λ: 220 nm Amax: ≤0.70
λ: 235 nm Amax: ≤0.50
λ: 250 nm Amax: ≤0.20
λ: 270 nm Amax: ≤0.10
λ: 295-400 nm Amax: ≤0.01

application(s)

food and beverages

SMILES string

C1COCCO1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Dioxane is widely used as a solvent for various organic compounds in industry. It is a non-biodegradable contaminant found in industrial effluents. Its oxidative degradation by using oxygen and ozone has been useful in converting it into biodegradable products, which can be easily removed by conventional methods.

Application

1,4-Dioxane may be employed as a solvent in the synthesis of N-(4-(prop-2-ynyloxy)phenyl)acetamide and 3-(4-(prop-2-ynyloxy)phenyl)-3,4-dihydro-2H-benzo[e][1,3]oxazine.
High-purity, multipurpose solvent, tested for suitability in:
  • HPLC
  • Spectrophotometry
  • Residual testing
  • LC-MS applications.

Reported as a solvent in:
  • HPLC-MS study of fat-soluble vitamins (FSVs)
  • GC-MS analysis of phthalates in workplace air

Suitable for HPLC, spectrophotometry, environmental testing

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Self-curable benzoxazine functional polybutadienes synthesized by click chemistry.
Kukut M, et al.
Designed Monomers and Polymers, 12(2), 167-176 (2009)
Oxidation and biodegradability enhancement of 1,4-dioxane using hydrogen peroxide and ozone.
C D Adams et al.
Environmental science & technology, 28(11), 1812-1818 (1994-10-01)
Christof Aellig et al.
ChemSusChem, 5(9), 1737-1742 (2012-07-05)
The dehydration of D-fructose to 5-hydroxymethylfurfural was studied under single-phase conditions in the low boiling solvent 1,4-dioxane at moderate temperatures in the presence of the solid acid-catalyst Amberlyst-15. The reaction was first examined and optimized under batch conditions, where it
Alkene trifluoromethylation coupled with C-C bond formation: construction of trifluoromethylated carbocycles and heterocycles.
Hiromichi Egami et al.
Angewandte Chemie (International ed. in English), 52(14), 4000-4003 (2013-02-27)
Kiyoaki Ishimoto et al.
Biomacromolecules, 13(11), 3757-3768 (2012-09-19)
For developing broader application of biobased polymers, graft copolymers and comb polymers having poly(lactic acid) (PLA) side chains have been synthesized by using a macromonomer technique. PLA macromonomers (MMm) having a methacryloyl polymerizable group with different PLA chain length with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service