Skip to Content
Merck
All Photos(3)

Key Documents

289116

Sigma-Aldrich

Carbon tetrachloride

anhydrous, ≥99.5%

Synonym(s):

Tetrachloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CCl4
CAS Number:
Molecular Weight:
153.82
Beilstein:
1098295
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
≥99.5%
bp:
76-77 °C (lit.)
vapor pressure:
143 mmHg ( 30 °C)
91 mmHg ( 20 °C)

grade

anhydrous

Quality Level

vapor density

5.32 (vs air)

vapor pressure

143 mmHg ( 30 °C)
91 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

impurities

<0.002% water
<0.005% water (100 mL)

refractive index

n20/D 1.460 (lit.)

bp

76-77 °C (lit.)

mp

−23 °C (lit.)

density

1.594 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)(Cl)Cl

InChI

1S/CCl4/c2-1(3,4)5

InChI key

VZGDMQKNWNREIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Carbon tetrachloride has been used in the carbon tetrachloride induced mouse model to study liver injury and regeneration.
Please view www.aldrich.com/epaods regarding the EPA′s request for application information of Ozone Depleting Substances

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Ozone 1 - Skin Sens. 1B - STOT RE 1 Inhalation

Target Organs

Liver,Kidney

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cytokine treatment optimises the immunotherapeutic effects of umbilical cord-derived MSC for treatment of inflammatory liver disease
de Witte SF, et al
Stem Cell Research & Therapy, 8(1), 140-140 (2017)
Mohammad Rafi et al.
Materials (Basel, Switzerland), 11(4) (2018-03-29)
Graphene oxide/poly(amidoamine) (GO/PAMAM) nanocomposite adsorbed high quantities of congo red (CR) anionic dye in 0.1 M NaCl solution, with the maximum adsorption capacity of 198 mg·g-1. The kinetics and thermodynamics of adsorption were investigated to elucidate the effects of pH
Long non-coding RNA PVT1 activates hepatic stellate cells through competitively binding microRNA-152
Zheng J, et al.
Oncotarget, 7(39), 62886-62886 (2016)
Mari Saif et al.
Nature biomedical engineering, 4(8), 801-813 (2020-06-24)
Monitoring the progression of non-alcoholic fatty liver disease is hindered by a lack of suitable non-invasive imaging methods. Here, we show that the endogenous pigment lipofuscin displays strong near-infrared and shortwave-infrared fluorescence when excited at 808 nm, enabling label-free imaging of
Keywan Mortezaee et al.
Canadian journal of physiology and pharmacology, 1-12 (2015-11-18)
Melatonin is known for being beneficial in targeting liver diseases. This study aimed to investigate whether melatonin post-treatment is capable of rat carbon tetrachloride (CCl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service