Skip to Content
Merck
All Photos(1)

Documents

299537

Sigma-Aldrich

Trifluoroacetic acid

≥99%, for protein sequencing

Synonym(s):

TFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COOH
CAS Number:
Molecular Weight:
114.02
Beilstein:
742035
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.9 (vs air)

Quality Level

vapor pressure

97.5 mmHg ( 20 °C)

type

for protein sequencing

Assay

≥99%

form

liquid

impurities

<0.05% water

evapn. residue

<0.005%

refractive index

n20/D 1.3 (lit.)

pH

1 (20 °C, 10 g/L)

bp

72.4 °C (lit.)

mp

−15.4 °C (lit.)

density

1.489 g/mL at 20 °C (lit.)

SMILES string

OC(C(F)(F)F)=O

InChI

1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

DTQVDTLACAAQTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trifluoroacetic acid is a naturally occurring organic solvent and biodegradable by microbial action. It can also be used as a catalyst for various organic transformations, including solvolysis, rearrangements, reductions, oxidations, and trifluoromethylation.

Application

Trifluoroacetic acid can be used as a solvent in the:
  • Mass spectrometry for protein identification.
  • Microwave-irradiated bromination of isoxazoles and pyrazoles with N-bromosuccinimide to synthesize mono-brominated isoxazoles and pyrazoles.
  • Lewis acid-catalyzed 6-endo-digcyclization of 2-(2-arylethynyl)heteroaryl esters to heterocyclic lactones.
  • Electrospinning of cellulose acetate to prepare the corresponding nanofibrous membranes.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Andrew C Tolonen et al.
Journal of visualized experiments : JoVE, (89)(89), doi:10-doi:10 (2014-07-22)
Stable isotope labeling of peptides by reductive dimethylation (ReDi labeling) is a method to accurately quantify protein expression differences between samples using mass spectrometry. ReDi labeling is performed using either regular (light) or deuterated (heavy) forms of formaldehyde and sodium
Continuous-flow synthesis of the anti-malaria drug artemisinin.
François Lévesque et al.
Angewandte Chemie (International ed. in English), 51(7), 1706-1709 (2012-01-18)
Macro- and micromethods for the determination of serum vitamin A using trifluoroacetic acid.
J B Neeld et al.
The Journal of nutrition, 79, 454-462 (1963-04-01)
Risto Kostiainen et al.
Journal of chromatography. A, 1216(4), 685-699 (2008-09-19)
The most widely used ionization techniques in liquid chromatography-mass spectrometry (LC-MS) are electrospray ionization (ESI), atmospheric pressure chemical ionization (APCI) and atmospheric pressure photoionization (APPI). All three provide user friendly coupling of LC to MS. Achieving optimal LC-MS conditions is
W C Mahoney et al.
The Journal of biological chemistry, 255(23), 11199-11203 (1980-12-10)
A method employing reverse phase high performance liquid chromatography has been developed for the purification of large denatured peptides. The cyanogen bromide fragments of hemoglobin alpha, beta, and gamma chains (13 to 91 residues in length) could be separated and

Protocols

BIOshell™ IgG 1000 Å C4 UHPLC Column for Improved Biomacromolecule Separations

BIOshell™ IgG 1000 Å C4 UHPLC Column for Improved Biomacromolecule Separations

BIOshell™ IgG 1000 Å C4 UHPLC Column for Improved Biomacromolecule Separations

BIOshell™ IgG 1000 Å C4 UHPLC Column for Improved Biomacromolecule Separations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service