Skip to Content
Merck
All Photos(3)

Key Documents

293245

Sigma-Aldrich

1-Octanol

suitable for HPLC, ≥99%

Synonym(s):

Alcohol C8, Capryl alcohol, Octyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OH
CAS Number:
Molecular Weight:
130.23
Beilstein:
1697461
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99%
technique(s):
HPLC: suitable
bp:
196 °C (lit.)
vapor pressure:
0.14 mmHg ( 25 °C)

vapor density

4.5 (vs air)

Quality Level

vapor pressure

0.14 mmHg ( 25 °C)

Assay

≥99%

form

liquid

autoignition temp.

523 °F

expl. lim.

8 %

technique(s)

HPLC: suitable

impurities

<0.010% water

evapn. residue

<0.004%

color

APHA: ≤10

refractive index

n20/D 1.429 (lit.)

bp

196 °C (lit.)

mp

−15 °C (lit.)

density

0.827 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 215 nm Amax: 1.00
λ: 225 nm Amax: 0.50
λ: 235 nm Amax: 0.20
λ: 250 nm Amax: 0.05
λ: 300-400 nm Amax: 0.01

SMILES string

CCCCCCCCO

InChI

1S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3

InChI key

KBPLFHHGFOOTCA-UHFFFAOYSA-N

Gene Information

mouse ... Chrna1(11435)

Looking for similar products? Visit Product Comparison Guide

General description

This solvent is glass distilled, submicron filtered and subjected to rigorous specification testing to provide lot-to-lot consistency.

Application

Designed for use with HPLC instrumentation and organic synthesis applications.
HPLC-grade 1-octanol is also suitable as a solvent/reagent for:
  • Solubility studies of clofazimine in drug formulation
  • Analysis of semi-volatile oxygenated compounds of pyrolysis bio oils by GC-MS

Biochem/physiol Actions

Blocks T-type Ca2+ channels

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

187.7 °F

Flash Point(C)

86.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 744-744 (1994)
Photocatalytic degradation of 1-octanol on anchored titanium oxide and on TiO2 powder catalysts.
Yamashita H, et al.
J. Catal., 158(1), 97-101 (1996)
Mechanisms of PVDF membrane formation by immersion-precipitation in soft (1-octanol) and harsh (water) nonsolvents.
Young TH, et al.
Polymer, 40(19), 5315-5323 (1999)
Characterization of titanium-silicon binary oxide catalysts prepared by the sol-gel method and their photocatalytic reactivity for the liquid-phase oxidation of 1-octanol.
Yamashita H, et al.
The Journal of Physical Chemistry B, 102(30), 5870-5875 (1998)
Cristian Gobbo et al.
ACS nano, 6(12), 10684-10698 (2012-12-12)
Controlling the molecular conformation of oligomers on surfaces through noncovalent interactions symbolizes an important approach in the bottom-up patterning of surfaces with nanoscale precision. Here we report on the design, synthesis, and scanning tunneling microscopy (STM) investigation of linear oligoamides

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service