Skip to Content
Merck
All Photos(1)

Documents

291773

Sigma-Aldrich

Sodium chloroacetate

98%

Synonym(s):

Chloroacetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2COONa
CAS Number:
Molecular Weight:
116.48
Beilstein:
3597157
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.55

Assay

98%

form

powder or chunks

mp

199 °C (dec.) (lit.)

SMILES string

[Na+].[O-]C(=O)CCl

InChI

1S/C2H3ClO2.Na/c3-1-2(4)5;/h1H2,(H,4,5);/q;+1/p-1

InChI key

FDRCDNZGSXJAFP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Nano-copper ions assembled cellulose-based composite with antibacterial activity for biodegradable personal protective mask.: This study explores the creation of a nano-copper ions cellulose-based composite, showcasing its potential antibacterial properties and application in biodegradable personal protective masks (Shao et al., 2023).

  • Effective extraction of fluoroquinolones from water using facile modified plant fibers.: This article investigates the use of modified plant fibers for the efficient extraction of fluoroquinolones from water, highlighting the utility of sodium chloroacetate in the modification process (Zhang et al., 2022).

  • Sodium Chloroacetate Modified Polyethyleneimine/Trimesic Acid Nanofiltration Membrane to Improve Antifouling Performance.: The study details the improvement of antifouling performance in nanofiltration membranes through modification with sodium chloroacetate (Gu et al., 2021).

  • Preparation of BiOCl/Bi(2)WO(6) Photocatalyst for Efficient Fixation on Cotton Fabric: Applications in UV Shielding and Self-Cleaning Performances.: This research focuses on the application of a BiOCl/Bi(2)WO(6) photocatalyst in UV shielding and self-cleaning performances on cotton fabric, employing sodium chloroacetate in the preparation process (Chen et al., 2021).

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

518.0 °F - closed cup

Flash Point(C)

270 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aliev et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 6(7), 1120-1126 (2000-04-29)
The thermally induced solidstate polymerization reactions in sodium chloroacetate and sodium bromoacetate, leading to poly(hydroxyacetic acid) (polyglycolide) and NaCl and NaBr, respectively, were studied by isothermal in situ solid-state NMR spectroscopy at 120, 130 and 140 degrees C with a
Omar A Al-Deeb et al.
Arzneimittel-Forschung, 56(1), 40-47 (2006-02-16)
The reaction of 3-(1-adamantyl)-4-substituted-1,2,4-triazoline-5-thiones 3a-g with sodium chloroacetate, in ethanolic sodium hydroxide yielded the corresponding N1-acetic acid derivatives 4a-g. The interaction of 3a-g with ethyl 2-bromopropionate in acetone, in the presence of potassium carbonate, yielded the corresponding N1-ethyl propionate derivatives
Removal of heavy metal ions from water by cross-linked carboxymethyl corn starch.
Kim BS and Lim S-T.
Carbohydrate Polymers, 39(3), 217-223 (1999)
Yirong Guo et al.
Food chemistry, 335, 127609-127609 (2020-08-03)
In this study, a fluorescence resonance energy transfer (FRET) immunoassay based on graphene oxide (GO) and up-converting nanoparticles (UCNPs) was established for rapid detection of imidacloprid, a commonly-used insecticide. Under 980 nm near-infrared light excitation, emission of UCNPs at 542 nm can
Kunkun Tu et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 7(7), 1902897-1902897 (2020-04-11)
The applicability of advanced composite materials with hierarchical structure that conjugate metal-organic frameworks (MOFs) with macroporous materials is commonly limited by their inferior mechanical properties. Here, a universal green synthesis method for the in situ growth of MOF nanocrystals within

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service