Skip to Content
Merck
All Photos(1)

Key Documents

276839

Sigma-Aldrich

Zinc chloride solution

1.0 M in diethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
form:
liquid
Pricing and availability is not currently available.

form

liquid

Quality Level

concentration

0.95-1.10 M (silver nitrate titration)
1.0 M in diethyl ether

density

0.835 g/mL at 25 °C

SMILES string

Cl[Zn]Cl

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Zinc chloride in combination with 1-ethyl-3-methylimidazolium chloride comprises a molten salt, which is employed in the electrodeposition studies of zinc on various substrates (glassy carbon and nickel).[1] ZnCl2 participates as a catalyst in the preparation of carbonyl-methylene condensation products[2] and poly(propylene fumarate).[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Albert K Shung et al.
Journal of biomaterials science. Polymer edition, 13(1), 95-108 (2002-05-11)
Diethyl fumarate and propylene glycol were reacted in the presence of a zinc chloride catalyst to synthesize poly(propylene fumarate) (PPF) over a period of 12 hours. The kinetics of the transesterification polymerization at 130 degrees C, 150 degrees C, and
Electrodeposition of zinc from a Lewis acidic zinc chloride-1-ethyl-3-methylimidazolium chloride molten salt.
Lin Y-F and Sun I.
Electrochimica Acta, 44(16), 2771-2777 (1999)
Zinc chloride as a new catalyst for Knoevenagel condensation.
Shanthan RP and Venkataratnam RV.
Tetrahedron Letters, 32(41), 5821-5822 (1991)
Kaushik Nath et al.
Environmental science and pollution research international, 20(6), 4030-4045 (2012-12-06)
Utilization of agrowaste materials for the production of activated carbon, as an excellent adsorbent with large surface area, is well established industrially, for dephenolation of wastewater. In the present work, dried pods of Prosopis cineraria-a novel and low-cost agrowaste material-were
İbrahim Gülseren et al.
Food chemistry, 135(2), 770-774 (2012-08-08)
Whey protein isolate (WPI) nanoparticles were prepared using ethanol desolvation, and their capacity to incorporate ZnCl(2) was analysed. Desolvation was carried out at pH 9 and the volume of added ethanol was 0-3 times the volume of protein solution. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service