Skip to Content
Merck
All Photos(3)

Key Documents

266426

Sigma-Aldrich

Iodine

ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
I2
CAS Number:
Molecular Weight:
253.81
Beilstein:
3587194
EC Number:
MDL number:
UNSPSC Code:
12141916
PubChem Substance ID:
NACRES:
NA.21

vapor density

9 (vs air)

Quality Level

vapor pressure

0.31 mmHg ( 25 °C)
1 mmHg ( 38.7 °C)

product line

ReagentPlus®

Assay

99.7% trace metals basis

form

beads

resistivity

1.3E15 μΩ-cm

particle size

1-3 mm

bp

184 °C (lit.)

mp

113 °C (lit.)

SMILES string

II

InChI

1S/I2/c1-2

InChI key

PNDPGZBMCMUPRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • High-confidence placement of low-occupancy fragments into electron density using the anomalous signal of sulfur and halogen atoms.: This study demonstrates the utilization of iodine′s anomalous scattering properties to enhance the accuracy of electron density maps in crystallography, providing crucial insights for chemists involved in molecular structure determination (Ma et al., 2024).

  • Aqueous Zinc-Iodine Pouch Cells with Long Cycling Life and Low Self-Discharge.: Research on zinc-iodine battery technology emphasizes iodine′s role in achieving low self-discharge rates and long life cycles, significant for developing more efficient energy storage systems (Wu et al., 2024).

  • Low-iodine-dose computed tomography coupled with an artificial intelligence-based contrast-boosting technique in children.: This innovative approach reduces iodine dosage in pediatric CT scans while maintaining image quality, crucial for minimizing radiation exposure and contrast-related risks (Shin et al., 2024).

  • Aqueous Electrolyte with Weak Hydrogen Bonds for Four-Electron Zinc-Iodine Battery Operates in a Wide Temperature Range.: Explores the electrochemical applications of iodine in battery technology, particularly in enhancing the performance of zinc-iodine batteries across diverse temperatures (Liu et al., 2024).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Respiratory system, Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Julien A Boos et al.
Nucleic acids research, 41(15), e145-e145 (2013-06-15)
Efficient tissue-specific delivery is a crucial factor in the successful development of therapeutic oligonucleotides. Screening for novel delivery methods with unique tissue-homing properties requires a rapid, sensitive, flexible and unbiased technique able to visualize the in vivo biodistribution of these
A spectrophotometric investigation of the interaction of iodine with aromatic hydrocarbons.
Benesi HA and Hildebrand JH.
Journal of the American Chemical Society, 71(8), 2703-2707 (1949)
Steven R Woodcock et al.
Free radical biology & medicine, 59, 14-26 (2012-12-04)
Nitrated fatty acids are the product of nitrogen dioxide reaction with unsaturated fatty acids. The discovery of peroxynitrite and peroxidase-induced nitration of biomolecules led to the initial reports of endogenous nitrated fatty acids. These species increase during ischemia/reperfusion, but concentrations
Tim I M Korevaar et al.
The Journal of clinical endocrinology and metabolism, 98(11), 4382-4390 (2013-09-17)
Premature delivery is an important risk factor for child mortality and psychiatric, metabolic, and cardiovascular disease later in life. In the majority of cases, the cause of prematurity cannot be identified. Currently, it remains controversial whether abnormal maternal thyroid function
Zulfiqar A Bhutta et al.
Lancet (London, England), 382(9890), 452-477 (2013-06-12)
Maternal undernutrition contributes to 800,000 neonatal deaths annually through small for gestational age births; stunting, wasting, and micronutrient deficiencies are estimated to underlie nearly 3·1 million child deaths annually. Progress has been made with many interventions implemented at scale and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service