Skip to Content
Merck
All Photos(3)

Key Documents

243388

Sigma-Aldrich

Magnesium oxide

ACS reagent, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgO
CAS Number:
Molecular Weight:
40.30
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.21
Assay:
97%
grade:
ACS reagent
form:
powder
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Assay

97%

form

powder

reaction suitability

reagent type: catalyst
core: magnesium

impurities

≤0.02% NH4OH pptd.
≤0.02% insol. dil. HCl
≤0.4% water soluble substances

loss

≤2.0% loss on ignition

mp

2852 °C (lit.)

anion traces

SO42- and SO32-: ≤0.02%
chloride (Cl-): ≤0.01%
nitrate (NO3-): ≤0.005%

cation traces

Ba: ≤0.005%
Ca: ≤0.05%
Fe: ≤0.01%
K: ≤0.005%
Mn: ≤5 ppm
Na: ≤0.5%
Sr: ≤0.005%
heavy metals (as Pb): ≤0.003%

SMILES string

O=[Mg]

InChI

1S/Mg.O

InChI key

CPLXHLVBOLITMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium oxide (MgO) is an industrially important high-temperature material.[1] Lithium-doped magnesium oxide (Li/MgO) may be employed for the synthesis of ethylene and ethane from methane.[2] It is widely employed as a catalyst support for carbon nanotubes since it possesses a high surface area, high temperature stability and easy removal by either acid or alkaline reagents.[3]

Application

Magnesium oxide (MgO) has been employed in the synthesis of bimetallic Co-Mo catalysts supported on different catalyst supports. These catalysts were used for single-walled carbon nanotube (SWCNT) synthesis.[4]
It may be employed as a phase stabilizer during the fabrication of porous β-tricalcium phosphate (β-TCP).[5]
Lithium-doped magnesium oxide (Li/MgO) may be employed for the synthesis of ethylene and ethane.[2] It may also be used as a catalyst in the preparation of (R)-phenyl carbonate.[3]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arantxa García-Legorreta et al.
Nutrients, 12(9) (2020-09-26)
Magnesium is a mineral that modulates several physiological processes. However, its relationship with intestinal microbiota has been scarcely studied. Therefore, this study aimed to assess the role of dietary magnesium content to modulate the intestinal microbiota of Wistar male rats.
Effect of different catalyst supports on the (n, m) selective growth of single-walled carbon nanotube from Co-Mo catalyst.
Wang B, et al.
J. Mater. Sci., 44(2), 3285-3295 (2009)
Synthesis of ethylene and ethane by partial oxidation of methane over lithium-doped magnesium oxide.
Ito T and Lunsford JH.
Nature, 721-722 (1985)
Fabrication of β-TCP foam: Effects of magnesium oxide as phase stabilizer on its properties.
Nikaido T, et al.
Ceramics International, 41(10), 14245-14250 (2015)
Magnesium oxide-catalysed reaction of carbon dioxide with an epoxide with retention of stereochemistry.
Yano T, et al.
Chemical Communications (Cambridge, England), 12, 1129-1130 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service