Skip to Content
Merck
All Photos(1)

Key Documents

24137

Sigma-Aldrich

2-Propanol

≥99.5% (GC), puriss., meets analytical specification of Ph. Eur., BP, USP

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.05
technique(s):
GC/GC: suitable
bp:
82 °C (lit.)
vapor pressure:
33 mmHg ( 20 °C)
44 mmHg ( 25 °C)
Pricing and availability is not currently available.

Product Name

2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)

vapor density

2.1 (vs air)

Quality Level

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

grade

puriss.

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

750 °F

quality

meets analytical specification of Ph. Eur., BP, USP

expl. lim.

2.0-12.7 %, 93 °C

technique(s)

GC/GC: suitable

impurities

acidity or alkalinity, complies
benzene and related subst., complies
peroxides, complies
residual solvents, complies
≤0.0001% benzene
≤0.0002% heavy metals (as Pb)
≤0.002% free acid (as C2H5COOH)
≤0.002% non-volatile matter
≤0.05% aldehydes and ketones (as CH3COCH3)
≤0.1% methanol (GC)
≤0.2% water (Karl Fischer)

refractive index

n20/D 1.377 (lit.)
n20/D 1.377-1.378

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

density

0.785 g/mL at 25 °C (lit.)

cation traces

Fe: ≤1 mg/kg

absorption

in accordance

suitability

complies for appearance of solution
complies for identity

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

The article number 24137-4X2.5L-R will be discontinued. Please order the single bottle 24137-2.5L-R which is physically identical with the same exact specifications.
The article number 24137-6X1L-R will be discontinued. Please order the single bottle 24137-1L-R which is physically identical with the same exact specifications.

greener alternative product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas W Winkler et al.
PLoS genetics, 11(10), e1005378-e1005378 (2015-10-02)
Genome-wide association studies (GWAS) have identified more than 100 genetic variants contributing to BMI, a measure of body size, or waist-to-hip ratio (adjusted for BMI, WHRadjBMI), a measure of body shape. Body size and shape change as people grow older
Emilie Montastier et al.
PLoS computational biology, 11(1), e1004047-e1004047 (2015-01-16)
Nutrigenomics investigates relationships between nutrients and all genome-encoded molecular entities. This holistic approach requires systems biology to scrutinize the effects of diet on tissue biology. To decipher the adipose tissue (AT) response to diet induced weight changes we focused on
Wei-Yu Lin et al.
Human molecular genetics, 24(1), 285-298 (2014-08-30)
Previous studies have suggested that polymorphisms in CASP8 on chromosome 2 are associated with breast cancer risk. To clarify the role of CASP8 in breast cancer susceptibility, we carried out dense genotyping of this region in the Breast Cancer Association
Swapna Ganapathy et al.
Journal of the American Chemical Society, 136(46), 16335-16344 (2014-10-24)
Fundamental research into the Li-O2 battery system has gone into high gear, gaining momentum because of its very high theoretical specific energy. Much progress has been made toward understanding the discharge mechanism, but the mechanism of the oxygen evolution reaction
Lin Jia et al.
Journal of biomaterials applications, 29(3), 364-377 (2014-04-01)
Fabricating scaffolds that can simulate the architecture and functionality of native extracellular matrix is a huge challenge in vascular tissue engineering. Various kinds of materials are engineered via nano-technological approaches to meet the current challenges in vascular tissue regeneration. During

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service