Skip to Content
Merck
All Photos(1)

Documents

238961

Sigma-Aldrich

Drierite

with indicator, 4 mesh

Synonym(s):

Calcium sulfate, Gypsum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaSO4
CAS Number:
Molecular Weight:
136.14
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

form

chunks

particle size

4 mesh

SMILES string

[Ca++].[O-]S([O-])(=O)=O

InChI

1S/Ca.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

OSGAYBCDTDRGGQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Drierite can be used as a desiccant in the isothiocyanate color test studies.

Legal Information

Drierite is a trademark of W.A. Hammond Drierite Co., LTD

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The ontogeny of personality in the wild guinea pig.
Guenther A, et al.
Animal behaviour, 90, 131-139 (2014)
Enthalpies of vaporization of organic compounds.
Howard PB and Wadso I.
Acta Chemica Scandinavica, 24, 145-149 (1970)
The analysis of carbon monoxide in urban air at the ppm level, and the normal carbon monoxide value.
L Dubois et al.
Journal of the Air Pollution Control Association, 16(3), 135-139 (1966-03-01)
Respirometry: anhydrous drierite equilibrates with carbon dioxide and increases washout times.
White CR, et al.
Physiological and Biochemical Zoology : Pbz, 79(5), 977-988 (2006)
Nathaniel Huebsch et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(27), 9762-9767 (2014-06-26)
Biological systems are exquisitely sensitive to the location and timing of physiologic cues and drugs. This spatiotemporal sensitivity presents opportunities for developing new therapeutic approaches. Polymer-based delivery systems are used extensively for attaining localized, sustained release of bioactive molecules. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service