Skip to Content
Merck
All Photos(1)

Key Documents

I19551

Sigma-Aldrich

Isoprene

Isoprene
1 of 1 reviewers received a sample product or took part in a promotion

99%, contains <1000 ppm p-tert-butylcatechol as inhibitor

Synonym(s):

2-Methyl-1,3-butadiene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHC(CH3)=CH2
CAS Number:
Molecular Weight:
68.12
Beilstein:
969158
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

2.35 (vs air)

Quality Level

vapor pressure

8.82 psi ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

428 °F

contains

<1000 ppm p-tert-butylcatechol as inhibitor

expl. lim.

10 %

refractive index

n20/D 1.422 (lit.)

bp

34 °C (lit.)

mp

−146 °C (lit.)

density

0.681 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C=C

InChI

1S/C5H8/c1-4-5(2)3/h4H,1-2H2,3H3

InChI key

RRHGJUQNOFWUDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Isoprene in the presence of sec-butyllithium as an initiator can undergo sequential anionic polymerization with styrene and methyl methacrylate to form polystyrene-block-polyisoprene-block-poly(methylmethacrylate) (SIM) and polyisoprene-block-polystyrene-block-poly(methyl methacrylate) (ISM), two asymmetric triblock terpolymers.[1] It is also used in the asymmetric synthesis of polystyrene-block-polyisoprene (PS-b-PI) diblock copolymers.[2]

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Flam. Liq. 1 - Muta. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-65.2 °F - closed cup

Flash Point(C)

-54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Four-Phase Morphologies in Blends of ABC and BAC Triblock Terpolymers
Haenelt TG, et al.
Macromolecular Chemistry and Physics, 219(1), 1700241-1700241 (2018)
Morphology and elasticity of polystyrene-block-polyisoprene diblock copolymers in the melt.
Haenelt TG, et al.
Korea-Australia Rheology Journal, 26(3), 263-275 (2014)
Is it useful to ask why plants emit isoprene?
Thomas D Sharkey
Plant, cell & environment, 36(3), 517-520 (2012-11-14)
Thomas D Sharkey et al.
Annals of botany, 101(1), 5-18 (2007-10-09)
Some, but not all, plants emit isoprene. Emission of the related monoterpenes is more universal among plants, but the amount of isoprene emitted from plants dominates the biosphere-atmosphere hydrocarbon exchange. The emission of isoprene from plants affects atmospheric chemistry. Isoprene
Tomohisa Kuzuyama et al.
Natural product reports, 20(2), 171-183 (2003-05-09)
This review covers the biosynthesis of the starter units of terpenoids, isopentenyl diphosphate (IPP) and dimethylallyl diphosphate (DMAPP) via the nonmevalonate pathway together with a new enzyme involved in the conversion of IPP and DMAPP, i.e type 2 IPP isomerase.

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

    • Reviews 3
    • Votes 9
    5 out of 5 stars.

    Good product

    Good product if you need isoprene. Was useful for atmospheric chemistry research.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service