Skip to Content
Merck
All Photos(3)

Key Documents

220345

Sigma-Aldrich

Sodium bromide

ReagentPlus®, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBr
CAS Number:
Molecular Weight:
102.89
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050404
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
form:
powder or crystals
solubility:
ethanol: soluble(lit.)
water: soluble(lit.)
Pricing and availability is not currently available.

grade

reagent

Quality Level

vapor pressure

1 mmHg ( 806 °C)

product line

ReagentPlus®

Assay

≥99%

form

powder or crystals

pH

5.4 (20 °C, 50 g/L)

mp

755 °C (lit.)

solubility

ethanol: soluble(lit.)
water: soluble(lit.)

SMILES string

[Na+].[Br-]

InChI

1S/BrH.Na/h1H;/q;+1/p-1

InChI key

JHJLBTNAGRQEKS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium bromide (NaBr) is an inorganic salt. It is an inexpensive source of bromide anion. It is used as anucleophilic catalyst in organic synthesis. Additionally, NaBr is a useful electrolyte in electrochemical oxidations.

Application

Sodium bromide is used:
  • As a catalyst for the microwavce assisted three-component cyclocondensation of aryl aldehydes, alkyl nitriles and dimedone for the synthesis of highly functionalized tetrahydrobenzo[b]pyrans.
  • As a source of bromine radical in the preparation of bromomethylated γ-lactones via electrochemical oxidative bromination and cyclization series of alkenoic acid derivatives.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium bromide catalysed one-pot synthesis of tetrahydrobenzo [b] pyrans via a three-component cyclocondensation under microwave irradiation and solvent free conditions.
Devi I and Bhuyan PJ.
Tetrahedron Letters, 45(47), 8625-8627 (2004)
Eagleson M.
Concise Encyclopedia Chemistry, 996-996 (1994)
Sergiu Coseri et al.
Carbohydrate polymers, 116, 9-17 (2014-12-03)
Unoxidized and carboxylated pullulan (obtained by pullulan oxidation using TEMPO-sodium hypochlorite-sodium bromide system) have been used as mediators for the silver nanoparticles formation (AgNPs), under environment-friendly conditions: using aqueous solutions, room temperature and notably, by using both mediators as reducing
Ling Yue et al.
Applied biochemistry and biotechnology, 175(7), 3557-3570 (2015-02-05)
A novel aqueous two-phase system (ATPS) only containing Gemini surfactant ethanediyl-1,2-bis(dodecyldimethylammonium bromide) and an inorganic salt sodium bromide was designed, and the physicochemical properties of the ATPS were investigated systematically. The results have shown that the coexisting two phases, one
Density, viscosity, and electrolytic conductivity of concentrated aqueous electrolyte solutions at several temperatures. Alkaline-earth chlorides, lanthanum chloride, sodium chloride, sodium nitrate, sodium bromide, potassium nitrate, potassium bromide, and cadmium nitrate.
Isono T.
Journal of Chemical and Engineering Data, 29(1), 45-52 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service