Skip to Content
Merck
All Photos(4)

Documents

215996

Sigma-Aldrich

Ammonium phosphate dibasic

ACS reagent, ≥98%

Synonym(s):

Ammonium hydrogenphosphate, Diammonium hydrogenphosphate, di-Ammonium hydrogenphosphate (sec)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2HPO4
CAS Number:
Molecular Weight:
132.06
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Assay

≥98%

form

crystals or chunks
powder

impurities

≤0.005% NH4OH pptd.
≤0.005% insolubles

pH

7.7-8.1 (25 °C, 5%)

mp

155 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.001%
nitrate (NO3-): ≤0.003%
sulfate (SO42-): ≤0.01%

cation traces

Ca: 0.001%
Fe: ≤0.001%
K: ≤0.005%
Mg: 0.0005%
Na: ≤0.005%
heavy metals (as Pb): ≤0.001%

SMILES string

N.N.OP(O)(O)=O

InChI

1S/2H3N.H3O4P/c;;1-5(2,3)4/h2*1H3;(H3,1,2,3,4)

InChI key

MNNHAPBLZZVQHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium phosphate dibasic is a water-soluble ammonium phosphate salt. It is a key component of phosphate fertilizers. It is commonly used as a flame retardant for plywood, paper, and fabrics, as well as a corrosion inhibitor consolidant for degraded carbonatic materials found in old buildings.

Application

Ammonium phosphate dibasic can be used as a catalyst:
  • To synthesize tetrahydrobenzo[b]pyranderivatives via three‐component condensation reaction.
  • In the solvent-free Biginelli reactions.
  • To synthesize bis(indolyl)methanes via solvent-free reaction of indole derivatives and aromatic or aliphatic aldehydes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diammonium hydrogenphosphate for the consolidation of building materials. Investigation of newly-formed calcium phosphates.
Elena P, et al.
Construction Building Materials, 195, 557-563 (2019)
Ash M and Ash I.
Handbook of Green Chemicals, 627-627 (2004)
Matteo Frasnelli et al.
Acta biomaterialia, 33, 283-289 (2016-01-23)
The beta to alpha transition in tricalcium phosphate (TCP) bioceramics containing different amount of magnesium was studied in the present work. Mg-doped TCP powder was obtained by solid-state reaction starting from pure calcium carbonate, ammonium phosphate dibasic and magnesium oxide
Electric Field-Assisted Solid Phase Extraction: Study of Electrochromatographic Parameters with an Anionic Model Compound.
Orlando RM, et al.
Journal of the Brazilian Chemical Society, 26(2), 310-318 (2015)
Study of metastable equilibria in the ternary system NH4H 2PO4-(NH4)2HPO4-H2O at -7 and -9 ?C.
Zerrouk L, et al.
Fluid Phase Equilibria, 354, 47-53 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service