Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium phosphate dibasic
Synonym(s):
Ammonium hydrogenphosphate, Diammonium hydrogenphosphate, di-Ammonium hydrogenphosphate (
sec
)
Linear Formula:
(NH
4
)
2
HPO
4
CAS Number:
7783-28-0
Molecular Weight:
132.06
EC Number:
231-987-8
Products
Product Category
salts (3)
bases (1)
buffers (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Purity
Quality Segment
Application
Special Grade
ACS reagent (2)
Puriss (1)
Puriss P.A. (1)
pharmaceutical primary standard (1)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
215996
Ammonium phosphate dibasic,
ACS reagent, ≥98%
Expand
View Pricing
A5764
Ammonium phosphate dibasic,
reagent grade, ≥98.0%
Expand
View Pricing
30402
Ammonium phosphate dibasic,
puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98% (alkalimetric)
Expand
View Pricing
379980
Ammonium phosphate dibasic,
≥99.99% trace metals basis
Expand
View Pricing
09839
Ammonium phosphate dibasic,
BioUltra, ≥99.0% (T)
Expand
View Pricing
1029986
Ammonium phosphate dibasic,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
79762
Ammonium phosphate dibasic,
suitable for HPLC, LiChropur
™
, 99.0-101.0% (T)
Expand
View Pricing