Skip to Content
Merck
All Photos(3)

Key Documents

158941

Sigma-Aldrich

Pentane

reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein:
969132
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
Assay:
98%
bp:
35-36 °C (lit.)
vapor pressure:
26.85 psi ( 55 °C)
8.43 psi ( 20 °C)

grade

reagent grade

Quality Level

vapor density

2.48 (vs air)

vapor pressure

26.85 psi ( 55 °C)
8.43 psi ( 20 °C)

Assay

98%

form

liquid

autoignition temp.

500 °F

expl. lim.

~8.3 %

refractive index

n20/D 1.358 (lit.)

bp

35-36 °C (lit.)

mp

−130 °C (lit.)

density

0.626 g/mL at 25 °C (lit.)

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentane (n-pentane) undergoes hydroisomerization in the presence of a nanocrystalline beta (BEA) zeolite. It isomerizes to iso-pentane in the presence of ZSM-5 (Zeolite Socony Mobil-5) zeolite catalysts under hydrogen and nitrogen atmosphere. Platinum catalysts when supported on sulfated zirconia-based solid superacids, promotes chemisorption and effective combustion of pentane.

Application

Pentane may be used for the fabrication of planar lipid bilayers on the films of amorphous PTFE (PTFE) by the “painting technique”.
Pentane may be used in the preparation of n-hexanal via photo-catalyzed carbonylation of the terminal methyl group in the presence of a rhodium catalyst.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytic combustion of n-pentane on Pt supported on solid superacids.
Hua W and Gao Z.
Applied Catalysis. B, Environmental, 17(1), 37-42 (1998)
Vivek Trivedi et al.
Pharmaceuticals (Basel, Switzerland), 12(4) (2019-10-17)
Solid core drug delivery systems (SCDDS) were prepared for the oral delivery of biomolecules using mesoporous silica as core, bovine haemoglobin (bHb) as model drug and supercritical fluid (SCF) processing as encapsulation technique. The use of organic solvents or harsh
Efficient catalytic C-H activation of alkanes: regioselective carbonylation of the terminal methyl group of n-pentane by RhCl (CO)(PMe 3) 2
Sakakura T and Tanaka M.
Journal of the Chemical Society. Chemical Communications, 10, 758-759 (1987)
Hydroisomerization of n-pentane over hybrid catalysts containing a supported hydrogenation catalyst.
Fujimoto K, et al.
Applied Catalysis A: General, 91(2), 81-86 (1992)
Hydroisomerization of pentane, hexane, and heptane for improving the octane number of gasoline.
Chica A and Corma A.
J. Catal., 187(1), 167-176 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service