Skip to Content
Merck
All Photos(1)

Key Documents

76870

Supelco

Pentane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein:
969132
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

2.48 (vs air)

Assay

≥99.5% (GC)

autoignition temp.

500 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

~8.3 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.358 (lit.)
n20/D 1.358

bp

35-36 °C (lit.)

mp

−130 °C (lit.)

density

0.626 g/mL at 25 °C (lit.)

application(s)

petroleum

format

neat

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pentane may be used as an analytical reference standard for the quantification of the analyte in human breath and ambient air samples using gas chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 4

1 of 4

A simple method for the trace determination of methanol, ethanol, acetone and pentane in human breath and in the ambient air by preconcentration on solid sorbents followed by gas chromatography
Qin T, et al.
Talanta, 44(9), 1683-1690 (1997)
Anna Svensson et al.
Carbohydrate polymers, 92(1), 775-783 (2012-12-12)
Dry ultra-porous cellulose fibres were obtained using a liquid exchange procedure in which water was replaced in the following order: water, methanol, acetone, and finally pentane; thereafter, the fibres were dried with Ar(g). The dry samples (of TEMPO-oxidized dissolving pulp)
Keisuke Maruyama et al.
Bioorganic & medicinal chemistry letters, 20(22), 6661-6666 (2010-10-05)
We have proposed a multi-template approach for drug development, focusing on similar fold structures of proteins, and have effectively generated lead compounds for several drug targets. Modification of these polypharmacological lead compounds is then needed to generate target-selective compounds. In
Dongsheng Guo et al.
Journal of hazardous materials, 186(2-3), 1788-1793 (2011-01-18)
The solvents n-pentane, methylene dichloride, ethyl ether and dodecylbenzenesulphonic acid sodium were used to regenerate exhausted activated carbon used in the process of treating coking wastewater, and the efficiency, ability, and optimum conditions of the different solvents on this regeneration
Fuming Chen et al.
Langmuir : the ACS journal of surfaces and colloids, 27(15), 9174-9181 (2011-07-01)
Graphene oxide (GO) can be viewed as an amphiphilic soft material, which form thin films at organic solvent-water interfaces. However, organic solvent evaporation provides little driving force, which results in slow GO transfer in aqueous phase, thus dawdling GO film

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service