Skip to Content
Merck
All Photos(1)

Key Documents

Y0001134

Raloxifene hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Keoxifene hydrochloride, LY 156758, [6-Hydroxy-2-(4-hydroxyphenyl)benzo[b]thien-3-yl][4-[2-(1-piperidinyl)ethoxy]phenyl]methanone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H27NO4S · HCl
CAS Number:
Molecular Weight:
510.04
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

raloxifene

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].Oc1ccc(cc1)-c2sc3cc(O)ccc3c2C(=O)c4ccc(OCCN5CCCCC5)cc4

InChI

1S/C28H27NO4S.ClH/c30-21-8-4-20(5-9-21)28-26(24-13-10-22(31)18-25(24)34-28)27(32)19-6-11-23(12-7-19)33-17-16-29-14-2-1-3-15-29;/h4-13,18,30-31H,1-3,14-17H2;1H

InChI key

BKXVVCILCIUCLG-UHFFFAOYSA-N

Gene Information

human ... ESR2(2100)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Raloxifene hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Raloxifene is a selective estrogen receptor modulator (SERM); acts as an anti-estrogen in both breast and uterine tissue while being estrogenic in bone. May have efficacy against estrogen-sensitive cancers.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pengcheng Luo et al.
Life sciences, 261, 118304-118304 (2020-08-24)
The signal transducer and activator of transcription 3 (STAT3) pathway plays an important role in inflammatory cascade process. Our previous studies found that Raloxifene targeted against IL-6/GP130 protein-protein interface and inhibited STAT3 phosphorylation induced by IL-6 in cancer cells. However
Tuan Hiep Tran et al.
Journal of nanoscience and nanotechnology, 14(7), 4820-4831 (2014-04-25)
The main aim of this study was to improve the oral bioavailability of raloxifene (RXF), a selective estrogen receptor modulator, by incorporation into solid lipid nanoparticles (SLN). RXF-loaded SLN was prepared by homogenization-sonication technique and characterized through physicochemical, pharmacokinetic, and
K R Snyder et al.
American journal of health-system pharmacy : AJHP : official journal of the American Society of Health-System Pharmacists, 57(18), 1669-1675 (2000-09-28)
The pharmacology, pharmacokinetics, clinical efficacy, adverse effects, and therapeutic role of raloxifene hydrochloride are reviewed. Raloxifene is a selective estrogen-receptor modulator (SERM) that has been approved for use in the prevention and treatment of osteoporosis in postmenopausal women. A SERM
[Raloxifene hydrochloride (Evista Tablet 60 mg) for postmenopausal osteoporosis: mode of action and clinical efficacy].
Hiroyuki Kobayashi et al.
Nihon yakurigaku zasshi. Folia pharmacologica Japonica, 125(1), 37-48 (2005-03-02)
Ibrahim A Abdelazim et al.
Archives of osteoporosis, 9, 189-189 (2014-07-20)
Osteoporosis is a major cause of mortality and morbidity worldwide. Decreased bone turnover markers and increased lumbar spine and total hip bone mineral density (BMD) in raloxifene-treated women add further support to the idea that raloxifene is an effective well-tolerated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service