Skip to Content
Merck
All Photos(1)

Key Documents

Y0000304

Arginine aspartate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

L-Arginine L-aspartate salt, S(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NO4 · C6H14N4O2
CAS Number:
Molecular Weight:
307.30
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

arginine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N[C@H](CCCN=C(N)N)C(=O)O.NC(CC(=O)O)C(=O)O

InChI

1S/C6H14N4O2.C4H7NO4/c7-4(5(11)12)2-1-3-10-6(8)9;5-2(4(8)9)1-3(6)7/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);2H,1,5H2,(H,6,7)(H,8,9)/t4-;/m1./s1

InChI key

SUUWYOYAXFUOLX-PGMHMLKASA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Arginine aspartate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bianca D Balmant et al.
Nutrition and cancer, 70(2), 249-256 (2018-01-19)
This study evaluated the effects of L-arginine supplementation on blood parameters, kidney and liver function, immunoglobulins and noninflammatory infiltrates in the small intestines of rats subjected to chemotherapy with 5-fluorouracil (5-FU). Thirty-two Wistar rats were randomly distributed into 4 groups
Peng Gao et al.
Journal of pharmaceutical and biomedical analysis, 118, 349-355 (2015-11-26)
Advanced adenomas are of higher risk to progress to colorectal cancer (CRC), the third leading cause of cancerous death worldwide. Endoscopy-based adenoma removal greatly contributes to arresting the progression of adenoma to CRC. Precise diagnosis, post-polypectomy surveillance and the follow-up

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service