Skip to Content
Merck
All Photos(2)

Key Documents

W360620

Sigma-Aldrich

D-(+)-Xylose

99%, natural sourced

Synonym(s):

Wood sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
FEMA Number:
3606
Beilstein:
1562108
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.21
grade:
Halal
Kosher
biological source:
corn
food allergen:
no known allergens

biological source

corn

Quality Level

grade

Halal
Kosher

reg. compliance

EU Regulation 178/2002

Assay

99%

mp

154-158 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

woody; smoky

SMILES string

O[C@@H]1COC(O)[C@H](O)[C@H]1O

InChI

1S/C5H10O5/c6-2-1-10-5(9)4(8)3(2)7/h2-9H,1H2/t2-,3+,4-,5?/m1/s1

InChI key

SRBFZHDQGSBBOR-IOVATXLUSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

D-(+)-Xylose is a pentose sugar first isolated from wood and found in coconuts, corn cobs, straw, and seed hulls . It is regarded as a safe food additive in the food industry to enhance the flavor and texture of various products such as cookies, fishery products, and bread.

Application

D-(+)-Xylose is used:
  • As an alternative sweetener in food products.
  • To produce the most common sugar alcohol, xylitol, which is used as a bulk sweetener in reduced-calorie foods.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lack of carcinogenicity of D-xylose given in the diet to F344 rats for two years
Kuroiwa Y, et al.
Food And Chemical Toxicology, 43(9), 1399-1404 (2005)
d-Xylose suppresses adipogenesis and regulates lipid metabolism genes in high-fat diet--induced obese mice
Lim E, et al.
Nutrition Research (New York, N.Y.), 35(7), 626-636 (2015)
Sugars alcohols: Chemical structures, manufacturing, properties and applications
Ibrahim OO
EC nutrition, 6, 817-824 (2016)
Lina M González et al.
Nature chemical biology, 16(2), 126-133 (2019-12-04)
Materials can be made multifunctional by embedding them with living cells that perform sensing, synthesis, energy production, and physical movement. A challenge is that the conditions needed for living cells are not conducive to materials processing and require continuous water

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service