Skip to Content
Merck
All Photos(1)

Key Documents

U0800000

Ursodeoxycholic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3α,7β-Dihydroxy-5β-cholan-24-oic acid, 5β-Cholan-24-oic acid-3α,7β-diol, 7β-Hydroxylithocholic acid, UDCS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H40O4
CAS Number:
Molecular Weight:
392.57
Beilstein:
3219888
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ursodeoxycholic acid

manufacturer/tradename

EDQM

mp

203-204 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16-,17-,18+,19+,20+,22+,23+,24-/m1/s1

InChI key

RUDATBOHQWOJDD-UZVSRGJWSA-N

Gene Information

human ... NR1H4(9971)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ursodeoxycholic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Carbone et al.
Gastroenterology, 144(3), 560-569 (2012-12-19)
BACKGROUND, & AIMS: Studies of primary biliary cirrhosis (PBC) phenotypes largely have been performed using small and selected populations. Study size has precluded investigation of important disease subgroups, such as men and young patients. We used a national patient cohort
Raoul Poupon
Clinics and research in hepatology and gastroenterology, 36 Suppl 1, S3-12 (2012-11-13)
Chronic cholestasis and liver inflammation are the two main pathophysiological components of the two major classes of disease - primary biliary cirrhosis (PBC) and primary sclerosing cholangitis (PSC) - leading to bile duct destruction and ultimately to cirrhosis and liver
Joana D Amaral et al.
Journal of lipid research, 50(9), 1721-1734 (2009-05-07)
Bile acids are a group of molecular species of acidic steroids with peculiar physical-chemical and biological characteristics. At high concentrations they become toxic to mammalian cells, and their presence is pertinent in the pathogenesis of several liver diseases and colon
Marcelo G Roma et al.
Clinical science (London, England : 1979), 121(12), 523-544 (2011-08-23)
UDCA (ursodeoxycholic acid) is the therapeutic agent most widely used for the treatment of cholestatic hepatopathies. Its use has expanded to other kinds of hepatic diseases, and even to extrahepatic ones. Such versatility is the result of its multiple mechanisms
Ursodeoxycholic acid for the treatment of primary biliary cirrhosis.
Keith Lindor
The New England journal of medicine, 357(15), 1524-1529 (2007-10-12)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service