Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ursodeoxycholic acid
Synonym(s):
3α,7β-Dihydroxy-5β-cholan-24-oic acid, 5β-Cholan-24-oic acid-3α,7β-diol, 7β-Hydroxylithocholic acid, UDCS
Empirical Formula (Hill Notation):
C
24
H
40
O
4
CAS Number:
128-13-2
Molecular Weight:
392.57
EC Number:
204-879-3
Products
Product Category
detergents (1)
sterols (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Package Size
API Family
Special Grade
pharmaceutical primary standard (2)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
U5127
Ursodeoxycholic acid,
≥99%
Expand
View Pricing
700199P
Ursodeoxycholic acid,
Avanti Research
™
- A Croda Brand
Expand
View Pricing
U0800000
Ursodeoxycholic acid,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing
BP706
Ursodeoxycholic acid,
British Pharmacopoeia (BP) Reference Standard
Expand
View Pricing