SYNPLE-B031
Reagent-cartridge Cbz Protection
Synonym(s):
Synple 2
About This Item
Recommended Products
description
Synple
SMILES string
[s]1cnc(c1c2ccc(cc2)CNC(=O)[C@H]3N(C[C@@H](C3)O)C(=O)[C@@H](NC(=O)CCOCCC(=O)O)C(C)(C)C)C
InChI key
HVPZHRHEHKZOOJ-TYBLODHISA-N
Looking for similar products? Visit Product Comparison Guide
General description
Scale: up to 0.5 mmol
Compatible substrates: carboxylic acids, primary and secondary alkyl amine saltsCartridge contents:
- Supported carbodiimide and Oxyma Pure (coupling reagents)
- SCX2 (for purification)
- Supported carbonate (acid scavenger)Learn more about the Synple automated synthesis platform.
Features and Benefits
- Time savings – pre-installed protocols have been highly optimized, which obviates the need for the user’s own reaction development and optimization
- Easy to use – protocols are loaded by simply scanning the cartridge; does not requires any programming knowledge
- Safety – all chemicals are contained in the system, minimizing the user’s exposure
- Set-up reaction in less than 5 minutes
- Small footprint
- Plug & Play system – simple installation
- Reaction temperatures from 25 °C – 80 °C
- Cartridges available for many different reactions
- Prefilled with all required reagents
- RFID chip containing optimized reaction protocol
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service