SYNPLE-H007
Reagent-cartridge 3-Methylmorpholine
Synple
Synonym(s):
Synple 2
About This Item
Recommended Products
Quality Level
parameter
up to 0.5 mmol scale
SMILES string
[s]1cnc(c1c2ccc(cc2)CNC(=O)[C@H]3N(C[C@@H](C3)O)C(=O)[C@@H](NC(=O)CCOCCC(=O)O)C(C)(C)C)C
InChI key
HVPZHRHEHKZOOJ-TYBLODHISA-N
Looking for similar products? Visit Product Comparison Guide
General description
Scale: up to 0.5 mmol
Heterocycle: 3-Methylmorpholine
Compatible substrates: aldehyde
Cartridge contents:
- Immobilized SnAP reagent
- Cu catalyst (Cu(OTF)2 and 2,6-lutidinium triflate)
- SCX (for purification)
- Silica (for purification)
Related Cartridges:
- SYNPLE-H001Reagent-cartridge Morpholine
- SYNPLE-H002Reagent-cartridge Oxazepane
- SYNPLE-H004Reagent-cartridge 2-Methylmorpholine
- SYNPLE-H005Reagent-cartridge Piperazine
- SYNPLE-H006Reagent-cartridge Diazepane
- SYNPLE-H010Reagent-cartridge Morpholine-2-spiro-(2-Pyr)
- SYNPLE-H011Reagent-cartridge Morpholine-2-spiro-(4-Pip)
- SYNPLE-H090Reagent-cartridge 9-OMe-Benzoxazepane
- SYNPLE-H091Reagent-cartridge 7-Br-9-OMe-Benzoxazepane
Learn more about the Synple automated synthesis platform.
Features and Benefits of Synple Platform
- Time savings - pre-installed protocols have been highly optimized, which obviates the need for the user′s own reaction development and optimization
- Easy to use - protocols are loaded by simply scanning the cartridge; does not requires any programming knowledge
- Safety - all chemicals are contained in the system, minimizing the user′s exposure
- Set-up reaction in less than 5 minutes
- Small footprint
- Plug & Play system - simple installation
- Reaction temperatures from 25 °C - 80 °C
- Cartridges available for many different reactions
- Prefilled with all required reagents
- RFID chip containing optimized reaction protocol
Other Notes
For more information on heterocycle formation using SnAP reagents, visit here: SnAP Technology Spotlight
Legal Information
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service