Skip to Content
Merck
All Photos(1)

Key Documents

SQC001

Supelco

Metals in Soil

certified reference material

Synonym(s):

Soil matrix CRM, heavy metals

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

manufacturer/tradename

RTC SQC-001

technique(s)

AAS: suitable
ICP: suitable

matrix

soil

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Signal Word

Danger

Hazard Statements

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rezvan Davodpour et al.
Archives of environmental contamination and toxicology, 77(3), 344-358 (2019-04-26)
Honeybees, due to their wide-ranging foraging behaviour, have great potentials for monitoring environmental quality. Therefore, the purpose of this study was to use honeybees as bioindicators for the detection of toxic and essential metals (Cd, Cr, Cu, and Fe) in
Xiaowen Ji et al.
Archives of environmental contamination and toxicology, 76(4), 540-553 (2019-02-24)
Permafrost-affected region in Russian Arctic is an important study area for investigating fate of trace metals in soils by geological processes and human-induced trace metals through atmospheric deposition. Two plots of soils in Yamal region were selected: Northern Trans-Urals area
Xiaowen Ji et al.
Journal of hazardous materials, 408, 124430-124430 (2020-11-13)
A 180-day incubation study was conducted to evaluate the effects of risk elements (REs) on organic carbon use and microbial activities in organic soils in the Arctic during the summer snowmelt period. Soils were artificially spiked with Cd, Pb, Cr

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service