Skip to Content
Merck
All Photos(1)

Key Documents

S4672

Sigma-Aldrich

Sodium sulfite

BioXtra, ≥98%

Synonym(s):

Natrium sulfite, Sodium sulphite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO3
CAS Number:
Molecular Weight:
126.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25
Assay:
≥98%
form:
powder or crystals
solubility:
H2O: 1 M at 20 °C, clear, colorless
Pricing and availability is not currently available.

product line

BioXtra

Quality Level

Assay

≥98%

form

powder or crystals

impurities

≤0.005% Insoluble matter
≤5 ppm Phosphorus (P)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤0.02%

cation traces

Al: ≤5 ppm
Ca: ≤0.01%
Cu: ≤5 ppm
Fe: ≤5 ppm
K: ≤0.05%
Mg: ≤0.005%
NH4+: ≤0.05%
Pb: ≤10 ppm
Zn: ≤5 ppm

SMILES string

[Na+].[Na+].[O-]S([O-])=O

InChI

1S/2Na.H2O3S/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2

InChI key

GEHJYWRUCIMESM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • A Soy Protein-Based Film Based on Chemical Treatment and Microcrystalline Cellulose Reinforcement Obtained from Corn Husk Byproducts.: This research explores the development of a soy protein-based film reinforced with microcrystalline cellulose derived from corn husk byproducts, treated chemically with sodium sulfite. The resulting film exhibited enhanced mechanical properties, making it a viable option for biodegradable packaging (Zhang et al., 2024) (Zhang et al., 2024).

  • Modelling Energy-Harvesting Processes in Primitive Cells: Proton Transport Across Bilayers Driven by the Oxidation of Sulfite.: This article models the energy-harvesting processes in primitive cells, focusing on proton transport across bilayers driven by sulfite oxidation. The findings contribute to the understanding of early cellular bioenergetics and the role of sulfite in these processes (Paula et al., 2024) (Paula et al., 2024).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amber R Boles et al.
Biotechnology and bioengineering, 109(3), 637-646 (2011-10-22)
A novel sulfur-utilizing perchlorate reducing bacterial consortium successfully treated perchlorate (ClO₄⁻) in prior batch and bench-scale packed bed reactor (PBR) studies. This study examined the scale up of this process for treatment of water from a ClO ₄⁻ and RDX contaminated
Michelle Cilia et al.
PloS one, 7(10), e48177-e48177 (2012-11-03)
Circulative transmission of viruses in the Luteoviridae, such as cereal yellow dwarf virus (CYDV), requires a series of precisely orchestrated interactions between virus, plant, and aphid proteins. Natural selection has favored these viruses to be retained in the phloem to
Qiang Yang et al.
Bioresource technology, 126, 336-340 (2012-11-06)
Agave americana is one of commonly grown agave species but currently less valuable because its large flower stalk cannot be used for producing alcoholic beverage. In the present study, the stalk was pretreated with dilute acid (DA), sulfite (SPORL), and
Jingang Wang et al.
Chemosphere, 91(3), 351-357 (2012-12-26)
This paper presents a simple, easy and reliable method for determination of EDTA (ethylenediaminetetraacetic acid) in aqueous system. Using EDTA chelating with ferric irons, an excessive amount of Fe(3+) was added to EDTA solution, and Fe(3+) in excess was reduced
Fabian Leinisch et al.
Chemical research in toxicology, 24(12), 2217-2226 (2011-10-19)
Nitrone spin traps such as 5,5-dimethyl-1-pyrroline-N-oxide (DMPO) are commonly used for free radical detection. Though proven examples are rare, artifact formation must be considered. For example, the Forrester-Hepburn mechanism yields the same radical adduct as that formed by genuine radical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service