Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium sulfite
Linear Formula:
Na
2
SO
3
CAS Number:
7757-83-7
Molecular Weight:
126.04
EC Number:
231-821-4
Products
Product Category
salts (9)
antioxidants (1)
preservatives (1)
Brand
SAFC (1)
Sigma-Aldrich (7)
Supelco (1)
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Anhydrous (3)
ACS reagent (1)
Puriss (1)
Puriss P.A. (1)
analytical standard (1)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
239321
Sodium sulfite,
ACS reagent, ≥98.0%
Expand
View Pricing
S0505
Sodium sulfite,
≥98%
Expand
View Pricing
1.06657
Sodium sulfite,
anhydrous for analysis EMSURE
®
Reag. Ph Eur
Expand
View Pricing
31454
Sodium sulfite,
puriss. p.a., reag. Ph. Eur., anhydrous, 98-100.0% (iodometric)
Expand
View Pricing
1.06652
Sodium sulfite,
EMPROVE
®
ESSENTIAL, Ph. Eur., BP, NF, E 221
Expand
View Pricing
71988
Sodium sulfite,
BioUltra, anhydrous, ≥98% (RT)
Expand
View Pricing
S4672
Sodium sulfite,
BioXtra, ≥98%
Expand
View Pricing
08981
Sodium sulfite,
analytical standard
Expand
View Pricing
901916
Sodium sulfite,
anhydrous, Redi-Dri
™
, reagent grade, ≥98%
Expand
View Pricing
13471
Sodium sulfite,
puriss., meets analytical specification of Ph. Eur., BP, NF, anhydrous, 95.0-100.0% (iodometric)
Expand
View Pricing