Skip to Content
Merck
All Photos(6)

Key Documents

R7649

Sigma-Aldrich

(−)-Riboflavin

meets USP testing specifications

Synonym(s):

Lactoflavin, Vitamin B2, Vitamin G

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N4O6
CAS Number:
Molecular Weight:
376.36
Beilstein:
97825
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058005
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

Assay

98.0-102.0% dry basis

form

powder

color

dark yellow

mp

290 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

CC1=C(C)C=C(N(C[C@H](O)[C@@H]([C@H](O)CO)O)C(C2=N3)=NC(NC2=O)=O)C3=C1

InChI

1S/C17H20N4O6/c1-7-3-9-10(4-8(7)2)21(5-11(23)14(25)12(24)6-22)15-13(18-9)16(26)20-17(27)19-15/h3-4,11-12,14,22-25H,5-6H2,1-2H3,(H,20,26,27)/t11-,12+,14-/m0/s1

InChI key

AUNGANRZJHBGPY-SCRDCRAPSA-N

General description

Riboflavin is an essential vitamin soluble in water. Milk and eggs are found to be its major sources. Riboflavin is usually bound to enzymes in all tissues. During riboflavin localization in plasma, it is bound to albumin and specific immunoglobulins.

Application

(-)-Riboflavin has been used to study its antimicrobial effect in vitro, through ultraviolet light A (UVA) activation.It has also been used collagen coated surface of the polyethylene membrane for cross-linking astrocytes.

Biochem/physiol Actions

Riboflavin serves as a precursor for the active enzyme cofactors riboflavin 5′-monophosphate (also called flavin mononucleotide or FMN) and flavin adenine dinucleotide (FAD). Riboflavin deficiency in the diet results in a well-defined syndrome known as ariboflavinosis, Riboflavin exhibits protective effects against tumor development and cardiovascular disease. Its deficiency often affects metabolism involving redox reactions. Riboflavin is found essential for iron absorption, gastrointestinal development, neurogenesis, corneal vascularization and corneal opacity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of antibacterial efficacy of photo-activated riboflavin using ultraviolet light (UVA)
Makdoumi K, et al.
Graefe'S Archive For Clinical and Experimental Ophthalmology = Albrecht Von Graefes Archiv Fur Klinische Und Experimentelle Ophthalmologie, 248(2), 207-212 (2010)
Photochemical eradication of methicillin-resistant Staphylococcus aureus by blue light activation of riboflavin
Makdoumi K, et al.
Acta Ophthalmologica, 95(5), 498-502 (2017)
Graciely Gomes Correa et al.
Data in brief, 31, 105736-105736 (2020-06-09)
Modular and tuneable genetic tools for Metabolic Engineering fuels the development of chassis for the efficient production of biocompounds at industrial scale. We have constructed an autoinduction device for gene expression in Bacillus subtilis based on the LuxR/I quorum sensing
Mechanical stretching elevates peptidyl arginine deiminase 2 expression in astrocytes
Algeciras ME, et al.
Current Eye Research, 33(11-12), 994-1001 (2008)
The efficiency of cross-linking methods in eradication of bacteria is influenced by the riboflavin concentration and the irradiation time of ultraviolet light
Backman A, et al.
Acta Ophthalmologica, 92(7), 656-661 (2014)

Articles

Riboflavin's impact on cell culture systems for manufacturing proteins and monoclonal antibodies discussed.

Riboflavin's impact on cell culture systems for manufacturing proteins and monoclonal antibodies discussed.

Riboflavin's impact on cell culture systems for manufacturing proteins and monoclonal antibodies discussed.

Riboflavin's impact on cell culture systems for manufacturing proteins and monoclonal antibodies discussed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service