Skip to Content
Merck
All Photos(1)

Key Documents

1.07609

Millipore

Riboflavine

for biochemistry

Synonym(s):

Riboflavine, Lactoflavine, Vitamin B2

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
376.36
MDL number:
UNSPSC Code:
12352205
EC Index Number:
201-507-1
NACRES:
NA.21

Quality Level

form

solid

potency

>10000 mg/kg LD50, oral (rat)

pH

5.5-7.2 (20 °C, 0.07 g/L in H2O)

mp

290 °C (decomposition)

solubility

0.07 g/L

bulk density

100 kg/m3

storage temp.

15-25°C

InChI

1S/C17H20N4O6/c1-7-3-9-10(4-8(7)2)21(5-11(23)14(25)12(24)6-22)15-13(18-9)16(26)20-17(27)19-15/h3-4,11-12,14,22-25H,5-6H2,1-2H3,(H,20,26,27)

InChI key

AUNGANRZJHBGPY-UHFFFAOYSA-N

General description

for biochemistry

Application

Riboflavine for biochemistry. CAS 83-88-5, pH 5.5 - 7.2 (0.07 g/l, H₂O, 20 °C).

Analysis Note

Assay (spectrophotometrically; calc. on dried substance)98.0 - 102.0 %Identity (IR-spectrum)conformsSpec. rotation [α²0/D (5 g/l, sodium hydroxide (0.05 mol/l), calc. on dried substance, Ph Eur)-135 - -115 °Lumiflavine (HPLC)conformsLoss on Drying (105 °C)≤ 1.5 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service