PHR1970
Acetaminophen Impurity C
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
N-(3-chloro-4-hydroxyphenyl)acetamide
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
form
solid
CofA
current certificate can be downloaded
packaging
pkg of 100 mg
application(s)
pharmaceutical (small molecule)
storage temp.
2-30°C
Related Categories
General description
Analysis Note
Other Notes
Footnote
Recommended products
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service