A7000
2-Acetamidophenol
97%
Synonym(s):
2′-Hydroxyacetanilide, N-(2-Hydroxyphenyl)acetamide, NSC 3989
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CONHC6H4OH
CAS Number:
Molecular Weight:
151.16
Beilstein:
607592
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
powder
mp
205-210 °C (lit.)
SMILES string
CC(=O)Nc1ccccc1O
InChI
1S/C8H9NO2/c1-6(10)9-7-4-2-3-5-8(7)11/h2-5,11H,1H3,(H,9,10)
InChI key
ADVGKWPZRIDURE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
P Anilkumar et al.
Langmuir : the ACS journal of surfaces and colloids, 24(17), 9754-9762 (2008-08-06)
Here, we report a synthesis of novel polyaniline nanospheres bearing mono- and bishydroxyl functional groups to trace the molecular interactions at the nanosurfaces through vitamin C sensing. Two new aniline monomers were synthesized via a tailor-made approach and polymerized to
K Pusecker et al.
The Journal of antibiotics, 50(6), 479-483 (1997-06-01)
The novel 5,10-dihydrophencomycin methyl ester (4) and the known microbial metabolites (2-hydroxyphenyl)-acetamide (1), menaquinone MK9 (II, III, VIII, IX-H8) (2), and phencomycin (3a) were isolated from an unidentified marine Streptomyces sp. and the structures were elucidated by NMR methods. Compound
Huma Jawed et al.
International immunopharmacology, 10(8), 900-905 (2010-05-11)
The present study was carried out to study the anti-arthritic and anti-inflammatory activity of N-(2-hydroxy phenyl) acetamide in adjuvant-induced arthritis in adult female Sprague Dawley rats. During experimental period, body weight and paw oedema volume were observed. At the end
Iryna Andrusenko et al.
Angewandte Chemie (International ed. in English), 58(32), 10919-10922 (2019-06-19)
Orthocetamol is a regioisomer of the well-known pain medication paracetamol and a promising analgesic and an anti-arthritic medicament itself. However, orthocetamol cannot be grown as single crystals suitable for X-ray diffraction, so its crystal structure has remained a mystery for
Kati Hanhineva et al.
The Journal of nutrition, 144(7), 1016-1022 (2014-05-09)
The bioavailability of whole-grain rye-derived phytochemicals has not yet been comprehensively characterized, and different baking and manufacturing processes can modulate the phytochemical composition of breads and other rye products. The aim of our study was to find key differences in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service