Skip to Content
Merck
All Photos(1)

Key Documents

PHR1678

Supelco

Bicalutamide

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Bicalutamide (CDX), Casodex, Cosudex, N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14F4N2O4S
CAS Number:
Molecular Weight:
430.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 1115
traceable to Ph. Eur. Y0001444
traceable to USP 1071202

API family

bicalutamide

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(O)(CS(=O)(=O)c1ccc(F)cc1)C(=O)Nc2ccc(C#N)c(c2)C(F)(F)F

InChI

1S/C18H14F4N2O4S/c1-17(26,10-29(27,28)14-6-3-12(19)4-7-14)16(25)24-13-5-2-11(9-23)15(8-13)18(20,21)22/h2-8,26H,10H2,1H3,(H,24,25)

InChI key

LKJPYSCBVHEWIU-UHFFFAOYSA-N

Gene Information

human ... AR(367)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Bicalutamide is a non-steroidal peripheral androgen receptor inhibitor. Its mode of action involves inhibiting androgen by binding to cytosol androgen receptors in the target tissue.

Application

Bicalutamide may be used as a pharmaceutical reference standard for the determination of the analyte by high performance liquid chromatography and UV spectrophotometry.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Bicalutamide (CDX) is a non-steriodal Androgen Receptor (AR) Antagonist and a pure antiandrogen.
Bicalutamide (CDX) is a non-steriodal Androgen Receptor (AR) antagonist and a pure antiandrogen. It acts via balancing histone acetylation/deacetylation and recruitment of coregulators. Bicalutamide (CDX) abolishes androgen-mediated expression. For example, MMP13 upregulation in prostate cancer, PLZF (promyelocytic leukemia zinc finger protein), and GADD45γ (growth arrest and DNA damage inducible, gamma). Bicalutamide (CDX) is inhibited by non-genomic, transcription-independent stimulation of PI3K/AKT phosphorylation by androgens.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA6123 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination and validation of UV spectrophotometric method for estimation of bicalutamide tablet
Swamivelmanickam M, et al,.
International Journal of ChemTech Research, 1(4), 1189-1193 (2009)
Development and validation of a stability indicating HPLC Method for determination of Bicalutamide in pharmaceutical formulations
Reddy GNK, et al.
International Journal of Pharmacy and Biological Sciences, 2(4), 134-149 (2012)
Development of an electrochemical method for the determination of bicalutamide at the SWCNT/CPE in pharmaceutical preparations and human biological fluids
Pandit UJ, et al.
Analytical Methods : Advancing Methods and Applications, 7(24), 10192-10198 (2015)
Development and validation of RP-HPLC method for the estimation of Bicalutamide in pure and pharmaceutical dosage forms
Rao AL, et al.
Rasayan Journal of Chemistry, 2(2), 512-515 (2009)
Emiliano Ventura et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 37(8), 1253-1263 (2020-05-14)
Selective androgen receptor modulators (SARMs) are a class of new emerging "designer" steroid compounds gaining popularity over more well established anabolic-androgenic steroids (AAS) amongst both non-professional and elite athletes. Moreover, due to their anabolic activity, SARM compounds may also potentially

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service